Key Documents
R8634
Sigma-Aldrich
Scienceware® centrifuge tube rack, epoxy-coated steel
20-position, for 15 mL plastic centrifuge tubes
Synonym(s):
wire tube holder
Select a Size
1 EA
€49.70
€49.70
Check Cart for Availability
Select a Size
Change View
1 EA
€49.70
About This Item
UNSPSC Code:
41122804
NACRES:
NB.35
€49.70
Check Cart for Availability
feature
Holds 20 x 18-20 mm tibes
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F18788-2003
size
12.5 cm × 11 cm × 6 cm
General description
  • Scienceware® centrifuge tube racks are made of epoxy-coated steel wire.
  • Polygrid racks are resistant to most of the chemicals including organic solvents, salts, weak acids, and weak alkalis.
  • Temperature range -223ºC (-369ºF) to 121ºC (250ºF); autoclavable.
Epoxy coated steel wire is chemical and corrosion resistant and steam autoclavable at 121ºC (250ºF).
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)