Skip to Content
Merck
All Photos(1)

Documents

R125

Sigma-Aldrich

Anti-RANTES antibody produced in goat

affinity isolated antibody, lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human

technique(s)

immunohistochemistry (frozen sections): suitable
neutralization: suitable
western blot: suitable

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CCL5(6352)

Related Categories

General description

RANTES (Regulated upon Activation, Normal T cell Expressed and Secreted; CCL5) is a chemokine that brings T cells, monocytes, eosinophils, NK cells, dendritic cells and mast cells to the site of infection or inflammation. RANTES has an unusual kinetic profile and is secreted 3-5 days after the activation of the T cells. It also exhibits antimicrobial activity by inducing the production of nitric oxide in macrophages. The main function of RANTES is mediated by CCR5, CCR1, CCR3 and other receptors. The expression of RANTES is regulated mainly by JAK/STAT and NF-κB pathways. Polymorphisms in the gene encoding RANTES are implicated in artherosclerosis, multiple sclerosis, HIV infection, diabetes, inflammatory diseases of the lung, renal disease and systemic lupus erythematosus. In addition, RANTES is a potential prognostic factor to track the progression of stage II breast cancer
Anti-RANTES antibody specifically recognizes human RANTES. The antibody shows ~40% cross-reactivity with mouse RANTES and canine RANTES, ~20% cross-reactivity with cotton rat RANTES, and ~1% cross-reactivity with feline RANTES.

Specificity

Based on ELISA, the antibody shows ~40% cross-reactivity with recombinant mouse RANTES, ~20% cross-reactivity with recombinant cotton rat RANTES, and ~1% cross-reactivity with recombinant feline RANTES.

Immunogen

recombinant human RANTES expressed in E. coli.

Application

Anti-RANTES antibody may be used for immunoblotting at a working concentration of 0.1 μg/ml. For immunohistochemistry and immunofluorescence, a working concentration of 5-15 μg/ml may be used. The antibody is suitable for neutralization reactions (ND50 is 0.1-0.4 μg/ml).

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Caitlin M Guenther et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 27(3), 611-622 (2019-02-18)
Adeno-associated virus (AAV) has emerged as a promising gene delivery vector because of its non-pathogenicity, simple structure and genome, and low immunogenicity compared to other viruses. However, its adoption as a safe and effective delivery vector for certain diseases relies
Neora Yaal-Hahoshen et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 12(15), 4474-4480 (2006-08-11)
The aim of this study was to determine the prognostic value of the chemokine CCL5, considered as a promalignancy factor in breast cancer, in predicting breast cancer progression and to evaluate its ability to strengthen the prognostic significance of other
V Appay et al.
Trends in immunology, 22(2), 83-87 (2001-04-05)
The activity of the chemokine RANTES is not restricted merely to chemotaxis. It is a powerful leukocyte activator, a feature potentially relevant in a range of inflammatory disorders. RANTES has attracted attention because it can potently suppress and, in some
Alan M Krensky et al.
Nature clinical practice. Nephrology, 3(3), 164-170 (2007-02-27)
Chemokines (chemoattractant cytokines) are fundamental regulators of immune cell movement from the bloodstream into tissues. Regulating expression of chemokines might, therefore, alleviate inflammation in autoimmune diseases and transplant rejection, or augment immune responses in cancer and immunodeficiency. RANTES (regulated upon
F Villalta et al.
Infection and immunity, 66(10), 4690-4695 (1998-09-24)
This paper describes a new role for the cysteine-cysteine (CC) chemokines RANTES, MIP-1alpha, and MIP-1beta on human macrophage function, which is the induction of nitric oxide (NO)-mediated trypanocidal activity. In a previous report, we showed that RANTES, MIP-1alpha and MIP-1beta

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service