Skip to Content
Merck
All Photos(1)

Key Documents

RNASEA-RO

Roche

RNase A

from bovine pancreas

Synonym(s):

Rnase

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€69.90
100 MG
€172.00

€69.90


Check Cart for Availability


Select a Size

Change View
25 MG
€69.90
100 MG
€172.00

About This Item

Enzyme Commission number:
UNSPSC Code:
12352204

€69.90


Check Cart for Availability

biological source

bovine pancreas

Quality Level

form

dry powder

specific activity

50 U/mg (+25°C, RNA as substrate)

mol wt

Mr = 13,700

packaging

pkg of 100 mg (10109169001)
pkg of 25 mg (10109142001)

manufacturer/tradename

Roche

parameter

65 °C optimum reaction temp.

technique(s)

flow cytometry: suitable

optimum pH

7.0-7.5

storage temp.

2-8°C

General description

Bovine pancreatic RNase A is a member of the RNase A protein superfamily. It is one of the most characterized proteins, and is a kidney shaped basic protein. This protein is composed of 124 amino acids. In its native form, RNase A exists as a homodimer.[1]

Application

  • For analytical purposes[2]
  • Isolation of DNA (for this purpose, RNase A should be boiled)[3]
  • For cell cycle analysis by flow cytometry and propidium iodide (PI) staining[4][5]

Biochem/physiol Actions

RNase A catalyzes the transphosphorylation and degradation of RNA. It was the first protein identified to exhibit DNA melting functionality. This enzyme binds to several binding sites on single stranded (ss) RNA polynucleotide chain before degrading it. It interacts with ssDNA in a similar manner, which is responsible for its ‘DNA unwinding′ property. RNase A dimer obtained through tandemization shows toxicity to cancer cells.[1]

Unit Definition

One unit is the enzyme activity that causes a decrease in absorbance of A0 to A1 within 1 minute under assay conditions. A0 to A1 corresponds to the total conversion, A1 being the final absorbance.

Reconstitution

RNase A can be dissolved at a concentration of 1 to 10 mg/ml in 10 mM Tris-HCl, pH 7.5, 15 mM NaCl, heated to 100 °C for 15 minutes to inactivate contaminating DNases and cooled slowly to room temperature and dispend into aliquots. Roche recommends subsequent storage at -15 to -25 °C.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amit Kumar Dinda et al.
Journal of photochemistry and photobiology. B, Biology, 162, 535-543 (2016-08-01)
Several DNA binding proteins exist in dimeric form when bound with DNA to be able to exhibit various biological processes such as DNA repair, DNA replication and gene expression. Various dimeric forms of Ribonuclease A (RNase A) and other members
Christopher J Ott et al.
Blood, 120(14), 2843-2852 (2012-08-21)
We investigated the therapeutic potential of JQ1, an inhibitor of the BET class of human bromodomain proteins, in B-cell acute lymphoblastic leukemia (B-ALL). We show that JQ1 potently reduces the viability of B-ALL cell lines with high-risk cytogenetics. Among the
Simon Asbjørn Larsen et al.
Chemistry Central journal, 6(1), 18-18 (2012-03-20)
Various by-products of the cellular metabolism, such as reactive carbonyl species (RCS) are potentially harmful to cells and tissues, and play a role in many physiological and pathological processes. Among various RCS is the highly reactive dicarbonyl glyoxal (GO), which
Bruno D Fonseca et al.
The Journal of biological chemistry, 290(26), 15996-16020 (2015-05-06)
The mammalian target of rapamycin complex 1 (mTORC1) is a critical regulator of protein synthesis. The best studied targets of mTORC1 in translation are the eukaryotic initiation factor-binding protein 1 (4E-BP1) and ribosomal protein S6 kinase 1 (S6K1). In this
Marieke Louwers et al.
Nature protocols, 4(8), 1216-1229 (2009-08-01)
Gene regulation in higher eukaryotes frequently involves physical interactions between genomic sequence elements tens of kilobases apart on the same chromosome but can also entail interactions between different chromosomes. Chromosome Conformation Capture (3C) is a powerful tool to identify such

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service