Skip to Content
Merck
All Photos(1)

Key Documents

Q2250

Sigma-Aldrich

3-Quinuclidone hydrochloride

Synonym(s):

1-Azabicyclo[2.2.2]octan-3-one hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H11NO · HCl
CAS Number:
Molecular Weight:
161.63
Beilstein:
3695039
EC Number:
MDL number:
UNSPSC Code:
12352200

mp

>300 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

Cl.O=C1CN2CC[C@H]1CC2

InChI

1S/C7H11NO.ClH/c9-7-5-8-3-1-6(7)2-4-8;/h6H,1-5H2;1H

InChI key

RFDPHKHXPMDJJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service