Skip to Content
Merck
All Photos(1)

Key Documents

Q1905

Sigma-Aldrich

3-Quinuclidone hydrochloride

97%

Synonym(s):

1-Azabicyclo[2.2.2]octan-3-one hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€84.50
100 G
€152.00

€84.50


Estimated to ship on28 March 2025


Request a Bulk Order

Select a Size

Change View
25 G
€84.50
100 G
€152.00

About This Item

Empirical Formula (Hill Notation):
C7H11NO · HCl
CAS Number:
Molecular Weight:
161.63
Beilstein:
3695039
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
97%

€84.50


Estimated to ship on28 March 2025


Request a Bulk Order

Quality Level

Assay

97%

mp

>300 °C (dec.) (lit.)

SMILES string

Cl.O=C1CN2CC[C@H]1CC2

InChI

1S/C7H11NO.ClH/c9-7-5-8-3-1-6(7)2-4-8;/h6H,1-5H2;1H

InChI key

RFDPHKHXPMDJJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antonio Zandona et al.
Archives of toxicology, 94(9), 3157-3171 (2020-06-26)
A library of 14 mono-oxime quinuclidinium-based compounds with alkyl or benzyl substituent were synthesized and characterized in vitro as potential antidotes for organophosphorus compounds (OP) poisoning treatment. We evaluated their potency for reversible inhibition and reactivation of OP inhibited human

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service