Skip to Content
Merck
All Photos(1)

Key Documents

PZ0369

Sigma-Aldrich

PF-04217329

≥98% (HPLC)

Synonym(s):

2-[3-[[[[4-(1H-Pyrazol-1-yl)phenyl]methyl](3-pyridinylsulfonyl)amino]methyl]phenoxy]-acetic acid 1-methylethyl ester, Isopropyl 2-[3-[[[[4-(1H-pyrazol-1-yl)phenyl]methyl](pyridin-3-ylsulfonyl)amino]methyl]phenoxy]acetate, PF 04217329, Taprenepag isopropyl

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€99.50
25 MG
€295.00

€99.50


Estimated to ship on17 May 2025



Select a Size

Change View
5 MG
€99.50
25 MG
€295.00

About This Item

Empirical Formula (Hill Notation):
C27H28N4O5S
CAS Number:
Molecular Weight:
520.60
UNSPSC Code:
41105101
NACRES:
NA.77

€99.50


Estimated to ship on17 May 2025


Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

room temp

SMILES string

O=S(N(CC1=CC=C(N2N=CC=C2)C=C1)CC3=CC=CC(OCC(OC(C)C)=O)=C3)(C4=CC=CN=C4)=O

InChI

1S/C27H28N4O5S/c1-21(2)36-27(32)20-35-25-7-3-6-23(16-25)19-30(37(33,34)26-8-4-13-28-17-26)18-22-9-11-24(12-10-22)31-15-5-14-29-31/h3-17,21H,18-20H2,1-2H3

InChI key

NVPXUFQLKWKBHK-UHFFFAOYSA-N

Biochem/physiol Actions

PF-04217329 is a selective and potent prostaglandin EP2 receptor agonist. PF-04217329 is an ocular bioavailable isopropyl ester prodrug of CP-544326 (active acid metabolite).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gina M Yanochko et al.
Journal of ocular pharmacology and therapeutics : the official journal of the Association for Ocular Pharmacology and Therapeutics, 30(5), 429-439 (2014-04-12)
Taprenepag isopropyl is an EP2 receptor agonist that is in development for the treatment of glaucoma. Iritis, photophobia, and increased corneal thickness observed in a Phase 2 clinical trial with taprenepag isopropyl were not previously observed in topical ocular toxicity
Ganesh Prasanna et al.
Experimental eye research, 93(3), 256-264 (2011-03-08)
Better control of intraocular pressure (IOP) is the most effective way to preserve visual field function in glaucomatous patients. While prostaglandin FP analogs are leading the therapeutic intervention for glaucoma, new target classes also are being identified with new lead

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service