Skip to Content
Merck
All Photos(1)

Key Documents

PZ0329

Sigma-Aldrich

ILS-920

≥98% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€69.40
25 MG
€300.00

€69.40


Estimated to ship on06 April 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€69.40
25 MG
€300.00

About This Item

Empirical Formula (Hill Notation):
C57H86N2O14
CAS Number:
Molecular Weight:
1023.30
UNSPSC Code:
12352200
NACRES:
NA.77

€69.40


Estimated to ship on06 April 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

shipped in

dry ice

storage temp.

−20°C

General description

Synonym: (3S,6R,7E,9R,10R,12R,14S,15S,18R,19E,21S,23S,26R,27R,34aS)-3,4,9,10,13,14,15,16,18,21,22,23,24,25,26,27,32,33,34,34a-eicosahydro-9,27-dihydroxy-3-[(1R)-2-[(1S,3R,4R)-4-hydroxy-3-methoxycyclohexyl]-1-methylethyl]-10,21-dimethoxy-6,8,12,14,20,26-hexamethyl-16-phenyl-23,27-epoxy-15,18-ethenopyrido[2,1-r][1,16,2,19]dioxadiazacyclohentriacontine-1,5,11,28,29(6H,12H,31H)-pentone

Biochem/physiol Actions

ILS-920 is a rapamycin derivative with reduced immunosuppressive activity and enhanced neuroprotective activities including efficacy in vivo stroke model. ILS-920 predominantly binds to FKBP52 with a 972-fold higher selectivity for FKBP52 vs. FKBP12 than that of rapamycin, and also binds to the β-subunit of L-type voltage dependent Ca2+ channels. This dual functionality may contribute to the compound′s efficacy in stroke models.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service