Skip to Content
Merck
All Photos(1)

Key Documents

PZ0297

Sigma-Aldrich

PF-06305591 dihydrate

≥98% (HPLC)

Synonym(s):

(αR,βS)-β-Amino-6-(1,1-dimethylethyl)-α-methyl-1H-benzimidazole-2-propanamide dihydrate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€128.00
25 MG
€517.00

€128.00


Estimated to ship on16 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€128.00
25 MG
€517.00

About This Item

Empirical Formula (Hill Notation):
C15H22N4O · 2H2O
CAS Number:
Molecular Weight:
310.39
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€128.00


Estimated to ship on16 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

shipped in

wet ice

storage temp.

−20°C

SMILES string

C[C@@H](C(N)=O)[C@H](N)C1=NC2=CC(C(C)(C)C)=CC=C2N1

InChI

1S/C15H22N4O/c1-8(13(17)20)12(16)14-18-10-6-5-9(15(2,3)4)7-11(10)19-14/h5-8,12H,16H2,1-4H3,(H2,17,20)(H,18,19)/t8-,12+/m1/s1

InChI key

APWZIFIAVVFPNT-PELKAZGASA-N

Biochem/physiol Actions

PF-06305591 is a potent and highly selective sodium channel Nav1.8 inhibitor.
PF-06305591 is safe and has acceptable absorption, distribution, metabolism, and elimination (ADME) properties.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alan D Brown et al.
Bioorganic & medicinal chemistry, 27(1), 230-239 (2018-12-13)
The voltage gated sodium channel NaV1.8 has been postulated to play a key role in the transmission of pain signals. Core hopping from our previously reported phenylimidazole leads has allowed the identification of a novel series of benzimidazole NaV1.8 blockers.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service