Skip to Content
Merck

Skip To

197331

Bcl-2 Inhibitor II, YC137

The Bcl-2 Inhibitor II, YC137, also referenced under CAS 810659-53-1, controls the biological activity of Bcl-2. This small molecule/inhibitor is primarily used for Activators/Inducers applications.

Synonym(s):

Bcl-2 Inhibitor II, YC137, 2-Methoxycarbonylamino-4-methylsulfanyl-butyric acid, 4-(4,9-dioxo-4,9-dihydronaphtho[2,3-d]thiazol-2-ylamino)-phenyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Check Cart for Availability
€ 223,00

About This Item

Empirical Formula (Hill Notation):
C24H21N3O6S2
CAS Number:
Molecular Weight:
511.57
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥90% (HPLC)
Form:
solid
Storage condition:
OK to freeze, protect from light

€ 223,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥90% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

red

solubility

DMSO: 5 mg/mL, ethanol: 5 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

[s]1c2c(nc1Nc4ccc(cc4)OC(=O)C(NC(=O)OC)CCSC)C(=O)c3c(cccc3)C2=O

InChI

1S/C24H21N3O6S2/c1-32-24(31)26-17(11-12-34-2)22(30)33-14-9-7-13(8-10-14)25-23-27-18-19(28)15-5-3-4-6-16(15)20(29)21(18)35-23/h3-10,17H,11-12H2,1-2H3,(H,25,27)(H,26,31)

InChI key

SFDSXIZFOOBDMC-UHFFFAOYSA-N

General description

A cell-permeable molecule that targets the BH3•domain interaction and selectively induces apoptosis in Bcl-2-overexpressing cells (<300 nM in MDA-MB435B breast cancer cells). Exhibits little effect on a variety of primary cells and Bcl-xL-dependent cells, even at a concentrations as high as 5 µM. Inhibits the binding of Bid BH3 to Bcl-2 thereby blocking the anti-apoptotic activity of Bcl-2.
A cell-permeable naphthoquinone compound that selectively induces apoptosis of Bcl-2-overexpressing cells (<300 nM in MDA-MB435B breast cancer cells) with little effect on a variety of primary cells, and Bcl-xL-dependent cells, even at a concentration of 5 µM. Preferentially binds Bcl-2 (Ki = 1.3 µM) and disrupts its interaction with Bid BH3, thereby blocking the anti-apoptotic activity of Bcl-2. Shown to induce the release of cytochrome c from mitochondria and activate caspase-9.

Biochem/physiol Actions

Cell permeable: yes
Product does not compete with ATP.
Reversible: no
Target Ki: 1.3 µM in binding Bcl-2

Packaging

Packaged under inert gas

Other Notes

Real, P.J., et al. 2004. Cancer Res. 64, 7947.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Carcinogenic / Teratogenic (D)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
220486197333238811
description

The Bcl-2 Inhibitor II, YC137, also referenced under CAS 810659-53-1, controls the biological activity of Bcl-2. This small molecule/inhibitor is primarily used for Activators/Inducers applications.

description

Chk2 Inhibitor II, BML-277, CAS 516480-79-8 is a cell-permeable, potent, reversible, ATP-competitive inhibitor of Chk2 (IC50 = 15 nM).

description

The Bcl-2 Inhibitor VI, ABT-737, also referenced under CAS 852808-04-9, controls the biological activity of Bcl-2. This small molecule/inhibitor is primarily used for Activators/Inducers applications.

description

The Cdk9 Inhibitor II, also referenced under CAS 140651-18-9, controls the biological activity of Cdk9. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

assay

≥90% (HPLC)

assay

≥95% (HPLC)

assay

≥97% (HPLC)

assay

≥95% (HPLC)

form

solid

form

solid

form

solid

form

solid

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage condition

OK to freeze, protect from light

storage condition

OK to freeze, protect from light

storage condition

OK to freeze, protect from light

storage condition

OK to freeze, protect from light


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters