Skip to Content
Merck
All Photos(1)

Key Documents

PZ0278

Sigma-Aldrich

PF-05020182

≥98% (HPLC)

Synonym(s):

N-[4,6-Dimethoxy-2-(4-methoxy-1-piperidinyl)-5-pyrimidinyl]-3,3-dimethyl-butanamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€95.20
25 MG
€372.00

€95.20


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€95.20
25 MG
€372.00

About This Item

Empirical Formula (Hill Notation):
C18H30N4O4
CAS Number:
Molecular Weight:
366.46
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€95.20


Estimated to ship on31 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

SMILES string

O=C(CC(C)(C)C)NC1=C(OC)N=C(N2CCC(OC)CC2)N=C1OC

InChI

1S/C18H30N4O4/c1-18(2,3)11-13(23)19-14-15(25-5)20-17(21-16(14)26-6)22-9-7-12(24-4)8-10-22/h12H,7-11H2,1-6H3,(H,19,23)

InChI key

XHJMCIALHJMEBB-UHFFFAOYSA-N

Biochem/physiol Actions

PF-05020182 is a Kv7.2/4 potassium channel opener.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service