189511
Autotaxin Inhibitor II, HA130
Synonym(s):
Autotaxin Inhibitor II, HA130, ATX Inhibitor II, (Z)-3-((4-((3-(4-Fluorobenzyl)-2,4-dioxo-1,3-thiazolan-5-yliden)-methyl)phenoxy)methyl)benzeneboronic acid
Select a Size
€176.00
Estimated to ship on26 May 2025
Select a Size
About This Item
€176.00
Estimated to ship on26 May 2025
Recommended Products
Assay
≥97% (HPLC)
Quality Level
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
desiccated (hygroscopic)
protect from light
color
off-white
solubility
DMSO: 50 mg/mL
shipped in
ambient
storage temp.
−20°C
General description
Packaging
Warning
Reconstitution
Other Notes
Albers, H.M.H.G. et al. 2010. Proc. Natl. Acad. Sci. USA107, 7257.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service