Skip to Content
Merck
All Photos(1)

Key Documents

PZ0220

Sigma-Aldrich

PF-06250112

≥98% (HPLC)

Synonym(s):

(R)-5-Amino-1-(1-cyanopiperidin-3-yl)-3-(4-[2,4-difluorophenoxy]phenyl)-1H-pyrazole-4-carboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€91.30
25 MG
€346.00

€91.30


Estimated to ship on20 May 2025



Select a Size

Change View
5 MG
€91.30
25 MG
€346.00

About This Item

Empirical Formula (Hill Notation):
C22H20F2N6O2
Molecular Weight:
438.43
UNSPSC Code:
12352200
NACRES:
NA.77

€91.30


Estimated to ship on20 May 2025


Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

−20°C

Biochem/physiol Actions

PF-06250112 is a potent inhibitor of Bruton′s tyrosine kinase (BTK).
PF-06250112 is a potent inhibitor of Bruton′s tyrosine kinase (BTK). PF-06250112 blocks both B-cell receptor and FcR-mediated signaling, and prevents FcR-dependent, Antibody-mediated proteinuria in a mouse glomerulonephritis model.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service