Skip to Content
Merck
All Photos(1)

Key Documents

219369

Sigma-Aldrich

Carcinoembryonic Antigen, Human Colon Adenocarcinoma Cell Line

Carcinoembryonic Antigen, Human Colon Adenocarcinoma Cell Line, is a native CEA from human colon adenocarcinoma cells.

Synonym(s):

CEA

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€664.00

€664.00


Estimated to ship on21 May 2025



Select a Size

Change View
100 μG
€664.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.42

€664.00


Estimated to ship on21 May 2025


Quality Level

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

aqueous buffer: soluble

shipped in

ambient

storage temp.

−20°C

General description

Native CEA from human colon adenocarcinoma cells. Found in tumors in adults. Widely used cancer marker found on many adult tumors. For more than a decade, purified CEA has been the most widely used general marker in determining and monitoring the status of malignant disease. This preparation is derived from an established adenocarcinoma cell line.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Analysis Note

single diffuse band by SDS-PAGE

Other Notes

Nomoto, K., et al. 1998. Jpn. J. Cancer Res. 89, 1257.
Jacobs, E.L., and Haskell, C.M. 1991. Curr. Probl. Cancer15, 299.
Robertson, J.F., et al. 1991. Br. J. Cancer64, 757.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lincidio Perez et al.
Biotechnology and applied biochemistry, 43(Pt 1), 39-48 (2005-08-17)
In the present paper we report the development of a bivalent scFv (single-chain Fv) antibody fragment, starting from a mouse mAb (monoclonal antibody) specific for CEA (carcinoembryonic antigen) that has received approval for in vivo radioimmunodiagnosis in humans. The diabody
Ana Alvarez-Cienfuegos et al.
Scientific reports, 6, 28643-28643 (2016-06-28)
Here, we describe a new strategy that allows the rapid and efficient engineering of mono and multispecific trivalent antibodies. By fusing single-domain antibodies from camelid heavy-chain-only immunoglobulins (VHHs) to the N-terminus of a human collagen XVIII trimerization domain (TIE(XVIII)) we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service