Skip to Content
Merck
All Photos(1)

Key Documents

PZ0158

Sigma-Aldrich

PF 3845 hydrate

≥98% (HPLC)

Synonym(s):

N-3-Pyridinyl-4-[[3-[[5-(trifluoromethyl)-2-pyridinyl]oxy]phenyl]methyl]-1-piperidinecarboxamide hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€124.00
25 MG
€481.00

€124.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€124.00
25 MG
€481.00

About This Item

Empirical Formula (Hill Notation):
C24H23F3N4O2 · xH2O
CAS Number:
Molecular Weight:
456.46 (anhydrous basis)
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

€124.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to tan

solubility

DMSO: ≥45 mg/mL

storage temp.

2-8°C

SMILES string

O.FC(F)(F)c1ccc(Oc2cccc(CC3CCN(CC3)C(=O)Nc4cccnc4)c2)nc1

InChI

1S/C24H23F3N4O2.H2O/c25-24(26,27)19-6-7-22(29-15-19)33-21-5-1-3-18(14-21)13-17-8-11-31(12-9-17)23(32)30-20-4-2-10-28-16-20;/h1-7,10,14-17H,8-9,11-13H2,(H,30,32);1H2

InChI key

CVQMBUCCSAKASE-UHFFFAOYSA-N

Biochem/physiol Actions

PF 3845 hydrate is a potent, irreversible FAAH inhibitor
PF 3845 is a potent, irreversible inhibitor of fatty acid amide hydrolase (FAAH), the principle enzyme involved in the degradation of the endocannabinoid anandamide. The endocannabinoid system is a target for therapeutic pain relief. PF-3845 is a covalent inhibitor that carbamylates FAAH′s serine nucleophile. It high selectivity over other enzymes including FAAH-2. In mouse studies, PF-3845 has been shown to raise brain anandamide levels for up to 24 hr; and in a rat model it produced significant reduction of inflammatory pain..

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service