Skip to Content
Merck
All Photos(1)

Key Documents

PZ0146

Sigma-Aldrich

SC-53116 hydrochloride hydrate

≥98% (HPLC)

Synonym(s):

4-Amino-5-chloro-N-[[(1S,7aS)-hexahydro-1H-pyrrolizin-1-yl]methyl]-2-methoxy-benzamide, SC 49518 hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H22ClN3O2 · HCl · xH2O
CAS Number:
Molecular Weight:
360.28 (anhydrous basis)
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

, white to yellow to brown

solubility

H2O: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

O.Cl.COc1cc(N)c(Cl)cc1C(=O)NC[C@@H]2CCN3CCCC23

InChI

1S/C16H22ClN3O2.ClH.H2O/c1-22-15-8-13(18)12(17)7-11(15)16(21)19-9-10-4-6-20-5-2-3-14(10)20;;/h7-8,10,14H,2-6,9,18H2,1H3,(H,19,21);1H;1H2/t10-,14-;;/m0../s1

InChI key

CZXMPKPHHWPFSI-UROSVOJDSA-N

Biochem/physiol Actions

SC-53116 is a 5-HT4 receptor partial agonist.
SC-53116 is a 5-HT4 receptor partial agonist. SC-53116 has been shown to ameliorate scopolamine-induced impairment in learning in rats at a does of 10 μg/rat i.c.v and increase population spike amplitude in the CA1 and CA3 fields of the hippocampus at the same concentration.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P G McLean et al.
British journal of pharmacology, 115(1), 47-56 (1995-05-01)
1. The pharmacological properties of 5-hydroxytryptamine (5-HT), the 5-HT4 receptor agonists, DAU 6236 and SC 53116 and the 5-HT4 receptor antagonist, GR 1130808, were studied in the rat oesophagus, rat ileum and human colon. 2. 5-HT relaxed the longitudinal muscle
L Oosterbosch et al.
Gut, 34(6), 794-798 (1993-06-01)
The aim was to investigate the action of serotonin (5HT) on function of the ileocolonic junction (ICJ) in vivo. In anaesthetised rats, models were developed to study the effects of intra-aortic (ia) serotonin on ileocolonic and colonic transit, and the
Daniel P Becker et al.
Bioorganic & medicinal chemistry letters, 14(12), 3073-3075 (2004-05-20)
Pyrrolizidine benzamide (+/-)-2, the bridgehead-methyl analog of SC-53116, was prepared and evaluated for 5-HT(4) agonism activity in the rat tunica muscularis (TMM) mucosae assay. Compound (+/-)-2 has an EC(50) of 449 nM in the TMM assay, as compared to 23
M Yamano et al.
Naunyn-Schmiedeberg's archives of pharmacology, 356(1), 145-150 (1997-07-01)
The influence of diabetic hyperglycemia on solid gastric emptying in rats was examined. Diabetes was produced by streptozotocin (STZ, 40 mg/kg i.v.), and diabetic hyperglycemia was observed from 1 day after the STZ injection. The gastric emptying of glass beads
S S Hegde et al.
Naunyn-Schmiedeberg's archives of pharmacology, 351(6), 589-595 (1995-06-01)
It is well documented that certain substituted benzamides, such as cisapride, and benzimidazolones, such as BIMU 8, enhance gastric emptying in rats. As these compounds possess 5-HT3 antagonistic and 5-HT4 agonistic properties, the precise mechanisms (5-HT3 or 5-HT4) underlying their

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service