Skip to Content
Merck
All Photos(1)

Key Documents

PZ0032

Sigma-Aldrich

PF-04479745

≥98% (HPLC)

Synonym(s):

(9S)-6,7,8,9-Tetrahydro-N,9-dimethyl-2-(phenylmethyl)-5H-pyrimido[4,5-d]azepin-4-amine, PF-4479745

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€92.70
25 MG
€282.00

€92.70


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€92.70
25 MG
€282.00

About This Item

Empirical Formula (Hill Notation):
C17H22N4
CAS Number:
Molecular Weight:
282.38
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

€92.70


Estimated to ship on28 May 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

SMILES string

CNC1=C2C([C@@H](C)CNCC2)=NC(CC3=CC=CC=C3)=N1

InChI

1S/C17H22N4/c1-12-11-19-9-8-14-16(12)20-15(21-17(14)18-2)10-13-6-4-3-5-7-13/h3-7,12,19H,8-11H2,1-2H3,(H,18,20,21)/t12-/m0/s1

InChI key

IHHALLDEDARSAL-LBPRGKRZSA-N

Biochem/physiol Actions

PF-04479745 (PF-4479745) is a selective serotonin 5HT2C agonist.
PF-04479745 is a selective serotonin 5HT2C agonist.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

DISCOVER Bioactive Small Molecules for Neuroscience

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service