Skip to Content
Merck
All Photos(3)

Key Documents

G8761

Sigma-Aldrich

(±)-Gossypol from cotton seeds

≥95% (HPLC), powder, PAF antagonist/inhibitor

Synonym(s):

(±)-2,2′-bis(8-Formyl-1,6,7-trihydroxy-5-isopropyl-3-methylnaphthalene)

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€274.00
250 MG
€561.00

€274.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 MG
€274.00
250 MG
€561.00

About This Item

Empirical Formula (Hill Notation):
C30H30O8
CAS Number:
Molecular Weight:
518.55
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€274.00


Estimated to ship on28 May 2025


Request a Bulk Order

Product Name

(±)-Gossypol from cotton seeds, ≥95% (HPLC)

Quality Level

Assay

≥95% (HPLC)

storage temp.

2-8°C

SMILES string

O=C([H])C1=C(C(O)=C(C2=C(C)C=C(C(C(C)C)=C(O)C(O)=C3C([H])=O)C3=C2O)C(C)=C4)C4=C(C(C)C)C(O)=C1O

InChI

1S/C30H30O8/c1-11(2)19-15-7-13(5)21(27(35)23(15)17(9-31)25(33)29(19)37)22-14(6)8-16-20(12(3)4)30(38)26(34)18(10-32)24(16)28(22)36/h7-12,33-38H,1-6H3

InChI key

QBKSWRVVCFFDOT-UHFFFAOYSA-N

General description

Gossypol is considered as an antifertility agent. It is present in cottonseed as a yellowish pigment. This reactive sesquiterpene aldehyde is present in the family Malvaceae and has a molecular mass of 518.54.[1]

Application

(±)-Gossypol from cotton seeds has been used:
  • as a standard to quantify the gossypol content in the extracts of leaves and flower buds[2]
  • to study its antifertility effects on spermatogenesis in NMRI mice[1]
  • to analyse its impact on human spermatozoa[3]
  • to incorporate into the artificial diet for larval rearing[4]

Biochem/physiol Actions

Gossypol is a male antifertility agent and PAF antagonist/inhibitor. Gossypol is a poisonous pigment found in cottonseed and its name is derived from the botanical name of the cotton plant, Gossypium.. Gossypium is extracted from selected hybrid species between two Gossypium species, Gossypium hirsutum and Gossypium barbadens. As a PAF antagonist/inhibitor, Gossypium markedly inhibited the contractile responses of guinea pig lung parenchyma strips stimulated with leukotriene B4, seukotriene D4, and PAF-acether.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Transcriptional responses underlying the hormetic and detrimental effects of the plant secondary metabolite gossypol on the generalist herbivore Helicoverpa armigera
de la Paz Celorio-Mancera M, et al.
BMC Genomics, 12(1), 575-575 (2011)
G Sarwar Gilani et al.
The British journal of nutrition, 108 Suppl 2, S315-S332 (2012-10-31)
Dietary antinutritional factors have been reported to adversely affect the digestibility of protein, bioavailability of amino acids and protein quality of foods. Published data on these negative effects of major dietary antinutritional factors are summarized in this manuscript. Digestibility and
Saiyi Zhong et al.
Anticancer research, 33(3), 949-955 (2013-03-14)
Breast cancer is the most commonly diagnosed cancer in women. Obesity is an important risk factor for developing breast cancer and is one of few risk factors that women can modify to prevent cancer. (-)-Gossypol-enriched cottonseed oil [(-)-GPCSO] contains 65%
Nariman Ansari et al.
Methods in cell biology, 113, 295-309 (2013-01-16)
Cell-based assays are essential in both basic research and drug discovery. Three-dimensional cellular spheroids are more realistic models of tumors and healthy tissues compared to standard two-dimensional cultures. Employing spheroids improves the reliability and the physiological significance of cell-based assays.
Carlos A Zelaya et al.
Acta crystallographica. Section C, Crystal structure communications, 69(Pt 4), 439-443 (2013-04-13)
Di(phenylpropylamino)gossypol [systematic name: 2,2'-bis{1,6-dihydroxy-5-isopropyl-8-[(3-phenylpropylamino)methylidene]naphthalen-7-one}, C48H52N2O6, was formed by reaction of the dimeric natural product gossypol with 3-phenylpropylamine. The structure of this compound has its two naphthalene ring systems oriented approximately perpendicular to each other, and the two pendant phenylpropyl groups

Articles

Warburg effect enhances glucose to lactate conversion in tumor cells, regardless of oxygen levels; impacting cancer metabolism since 1924.

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service