Skip to Content
Merck
All Photos(3)

Key Documents

PRS2513

Sigma-Aldrich

Anti-NOD2 (ab2) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-CARD15, Anti-Caspase recruitment domain 15, Anti-Nucleotide-binding oligomerization domain containing 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€413.00

€413.00


Estimated to ship on16 April 2025



Select a Size

Change View
100 μG
€413.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€413.00


Estimated to ship on16 April 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 114 kDa

species reactivity

human

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NOD2(64127)

Immunogen

a synthetic peptide corresponding to 14 amino acids at the carboxy-terminus of human NOD2.

Linkage

The action of this antibody can be blocked using blocking peptide SBP2513.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service