Skip to Content
Merck
All Photos(1)

Key Documents

P9864

Sigma-Aldrich

PTP-β human

≥90%, recombinant, expressed in E. coli K12, GST-tagged, buffered aqueous glycerol solution, ≥50,000 units/mg protein

Synonym(s):

Protein Tyrosine Phosphatase-β

Sign Into View Organizational & Contract Pricing

Select a Size

10000 UNITS
€625.00

€625.00


Estimated to ship on14 May 2025


Request a Bulk Order

Select a Size

Change View
10000 UNITS
€625.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

€625.00


Estimated to ship on14 May 2025


Request a Bulk Order

biological source

human

Quality Level

recombinant

expressed in E. coli K12

Assay

≥90%

form

buffered aqueous glycerol solution

specific activity

≥50,000 units/mg protein

mol wt

~250 kDa

UniProt accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PTPRB(5787)

General description

Protein tyrosine phosphatase, receptor type B (PTPRB), also known as vascular endothelial-protein-tyrosine phosphatase (VE-PTP), might have a role in remodeling of blood vessels and in angiogenesis. The gene encoding it is localized on human chromosome 12q15.[1] It is also important for endothelial cell migration.[2] The protein also controls the phosphorylation state of the angiopoietin receptor, Tie-2. PTPRB may modulate sodium channels by influencing the tyrosine phosphorylation status.[3][4]

Biochem/physiol Actions

Dephosphorylates tyrosine proteins.

Unit Definition

One unit of PTP-β will hydrolyze 1.0 nanomole of p-nitrophenyl phosphate per min at 30 °C.

Physical form

solution in 40 mM Tris-HCl, pH 8.0, 110 mM NaCl, 2.2 mM KCL, 3 mM DTT, 16 mM glutathione and 20% glycerol

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prioritization of Schizophrenia Risk Genes
by a Network-Regularized Logistic
Regression Method
Wen Zhang
INTERNATIONAL CONFERENCE OF COMPUTATIONAL METHODS IN SCIENCES AND ENGINEERING 2009:(ICCMSE 2009). (2016)
Picomolar concentrations of free zinc(II) ions regulate receptor protein-tyrosine phosphatase ? activity.
Wilson M
The Journal of Biological Chemistry (2012)
1002 protein phosphatases?
H Charbonneau et al.
Annual review of cell biology, 8, 463-493 (1992-01-01)
Shear stress-induced redistribution of vascular endothelial-protein-tyrosine phosphatase (VE-PTP) in endothelial cells and its role in cell elongation.
Mantilidewi KI
The Journal of Biological Chemistry (2014)
A sodium channel signaling complex: modulation by associated receptor protein tyrosine phosphatase beta.
Ratcliffe CF
Nature Neuroscience (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service