Skip to Content
Merck
All Photos(1)

Key Documents

GE27-1542-01

E. coli BL21

Cytiva 27-1542-01, pack of 1 vial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106501
NACRES:
NA.25
Pricing and availability is not currently available.

packaging

pack of 1 vial

manufacturer/tradename

Cytiva 27-1542-01

shipped in

dry ice

storage temp.

−20°C

General description

It is an exemplary host for the expression of recombinant proteins. As an E. coli B strain, it further lacks the major protease, encoded by the lon gene, catalyzing the endoproteolytic cleavage of damaged and recombinant proteins in the cell.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Storage Class Code

13 - Non Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service