Skip to Content
Merck
All Photos(1)

Key Documents

P5881

Sigma-Aldrich

Piperidine

≥99.0% (GC)

Synonym(s):

Hexahydropyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11N
CAS Number:
Molecular Weight:
85.15
Beilstein:
102438
EC Number:
MDL number:
UNSPSC Code:
12352103

vapor density

3 (vs air)

vapor pressure

23 mmHg ( 20 °C)

Assay

≥99.0% (GC)

refractive index

n20/D 1.452 (lit.)

bp

106 °C (lit.)

mp

−13 °C (lit.)

density

0.862 g/mL at 20 °C (lit.)

SMILES string

C1CCNCC1

InChI

1S/C5H11N/c1-2-4-6-5-3-1/h6H,1-5H2

InChI key

NQRYJNQNLNOLGT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

60.8 °F - closed cup

Flash Point(C)

16 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marek Łuczkowski et al.
Inorganic chemistry, 47(23), 10875-10888 (2008-10-31)
A de novo protein design strategy provides a powerful tool to elucidate how heavy metals interact with proteins.Cysteine derivatives of the TRI peptide family (Ac-G(LKALEEK)4G-NH2) have been shown to bind heavy metals in an unusual trigonal geometry. Our present objective
Total synthesis of (-)-kopsinine by an asymmetric one-pot [n+2+3] cyclization.
Shingo Harada et al.
Chemistry, an Asian journal, 7(10), 2196-2198 (2012-08-22)
J Phillip Turner et al.
ACS chemical neuroscience, 5(7), 552-558 (2014-04-03)
Alzheimer's disease (AD) is the most common form of dementia and the sixth leading cause of death in the United States. Plaques composed of aggregated amyloid-beta protein (Aβ) accumulate between the neural cells in the brain and are associated with
Xuwang Chen et al.
Bioorganic & medicinal chemistry, 20(12), 3856-3864 (2012-05-18)
A novel series of piperidine-linked amino-triazine derivatives were designed, synthesized and evaluated for in vitro anti-HIV activity as non-nucleoside reverse transcriptase inhibitors on the basis of our previous work. Screening results indicated that most compounds showed excellent activity against wild-type
Niina Aaltonen et al.
Chemistry & biology, 20(3), 379-390 (2013-03-26)
Monoacylglycerol lipase (MAGL) terminates the signaling function of the endocannabinoid, 2-arachidonoylglycerol (2-AG). During 2-AG hydrolysis, MAGL liberates arachidonic acid, feeding the principal substrate for the neuroinflammatory prostaglandins. In cancer cells, MAGL redirects lipid stores toward protumorigenic signaling lipids. Thus MAGL

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service