Merck
  • Piperazine and piperidine triazole ureas as ultrapotent and highly selective inhibitors of monoacylglycerol lipase.
Piperazine and piperidine triazole ureas as ultrapotent and highly selective inhibitors of monoacylglycerol lipase.
Chemistry & biology (2013-03-26)
Niina Aaltonen, Juha R Savinainen, Casandra Riera Ribas, Jani Rönkkö, Anne Kuusisto, Jani Korhonen, Dina Navia-Paldanius, Jukka Häyrinen, Piia Takabe, Heikki Käsnänen, Tatu Pantsar, Tuomo Laitinen, Marko Lehtonen, Sanna Pasonen-Seppänen, Antti Poso, Tapio Nevalainen, Jarmo T Laitinen
ABSTRACT
Monoacylglycerol lipase (MAGL) terminates the signaling function of the endocannabinoid, 2-arachidonoylglycerol (2-AG). During 2-AG hydrolysis, MAGL liberates arachidonic acid, feeding the principal substrate for the neuroinflammatory prostaglandins. In cancer cells, MAGL redirects lipid stores toward protumorigenic signaling lipids. Thus MAGL inhibitors may have great therapeutic potential. Although potent and increasingly selective MAGL inhibitors have been described, their number is still limited. Here, we have characterized piperazine and piperidine triazole ureas that combine the high potency attributable to the triazole leaving group together with the bulky aromatic benzodioxolyl moiety required for selectivity, culminating in compound JJKK-048 that potently (IC50 < 0.4 nM) inhibited human and rodent MAGL. JJKK-048 displayed low cross-reactivity with other endocannabinoid targets. Activity-based protein profiling of mouse brain and human melanoma cell proteomes suggested high specificity also among the metabolic serine hydrolases.
Product Description
Supelco
Piperazine hexahydrate, analytical standard
Sigma-Aldrich
Piperidine, ReagentPlus®, 99%
Sigma-Aldrich
Piperidine, ≥99.5%, purified by redistillation
Sigma-Aldrich
Piperidine, biotech. grade, ≥99.5%
Sigma-Aldrich
JJKK-048, ≥98% (HPLC)
Sigma-Aldrich
Piperazine, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Piperidine solution, suitable for peptide synthesis, 20% in DMF
Sigma-Aldrich
Piperazine, ReagentPlus®, 99%