Skip to Content
Merck
All Photos(1)

Key Documents

P3953

Sigma-Aldrich

L-α-Phosphatidyl-D-myo-inositol 3-monophosphate, dipalmitoyl

Sign Into View Organizational & Contract Pricing

Select a Size

0.1 MG
€287.00

€287.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
0.1 MG
€287.00

About This Item

Empirical Formula (Hill Notation):
C41H80O16P2
CAS Number:
Molecular Weight:
891.01
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.77

€287.00


Estimated to ship on26 May 2025


Request a Bulk Order

solubility

DMSO: soluble
chloroform: soluble
ethanol: soluble

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCC(=O)OC[C@H](COP(O)(=O)O[C@H]1[C@H](O)[C@@H](O)[C@H](O)[C@@H](OP(O)(O)=O)[C@H]1O)OC(=O)CCCCCCCCCCCCCCC

InChI

1S/C41H80O16P2/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-34(42)53-31-33(55-35(43)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2)32-54-59(51,52)57-41-38(46)36(44)37(45)40(39(41)47)56-58(48,49)50/h33,36-41,44-47H,3-32H2,1-2H3,(H,51,52)(H2,48,49,50)/t33-,36+,37+,38-,39-,40-,41+/m1/s1

InChI key

SZPQTEWIRPXBTC-KFOWTEFUSA-N

Substrates

Component of lipid signaling pathway; product of phosphatidylinositol 3-kinase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C L Huang et al.
Nature, 391(6669), 803-806 (1998-03-05)
Inward rectifier K+ channels, which modulate electrical activity in many cell types, are regulated by protein kinases, guanine-nucleotide-binding proteins (G proteins) and probably actin cytoskeleton. Generation of phosphatidylinositol 4,5-bisphosphate (PIP2) by ATP-dependent lipid kinases is known to activate inward rectifier
J E Harlan et al.
Biochemistry, 34(31), 9859-9864 (1995-08-08)
The pleckstrin homology (PH) domain is a protein module of approximately 100 amino acids that is found in several proteins involved in signal transduction [for a recent review, see Gibson et al. (1994) Trends Biochem. Sci. 19, 349-353]. Although the
J A Pitcher et al.
The Journal of biological chemistry, 270(20), 11707-11710 (1995-05-19)
The pleckstrin homology (PH) domain is an approximately 100-amino-acid region of sequence homology present in numerous proteins of diverse functions, which forms a discrete structural module. Several ligands capable of binding to PH domain-containing proteins have been identified including phosphatidylinositol

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service