Skip to Content
Merck
All Photos(1)

Documents

P4304

Supelco

Sigma Pseudo Corpse Scent

Formulation I

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

For early detection, or below 0 °C

Canine training aids for the detection of corpses.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Sigma pseudo corpse scent has been used as a test material for determining the components present in it using techniques such as a direct injection, headspace solid phase microextraction (SPME), purge and trap, and thermal desorption followed by gas chromatography coupled with mass spectrometry (GC/MS).
Canine training aids for the detection of corpses.

Legal Information

Sigma Pseudo is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comprehensive characterization of commercially available canine training aids
Tipple CA, et al.
Forensic Science International, 242, 242-254 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service