Merck
Skip To
P3049
Finnpipette® pipette tips
Finntip 250 Universal, non-sterile
Synonym(s):
Disposable pipette tips, Finnpipette tips
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Size/SKUAvailabilityPrice
1000 ea
Check Cart for Availability
€ 74,80
About This Item
NACRES:
NB.01
UNSPSC Code:
41121600
Sterility:
non-sterile
Feature:
barrier: no
Packaging:
pack of 1000 ea (Bag)
Manufacturer/tradename:
Finn 9400230
€ 74,80
Check Cart for Availability
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
colorless
sterility
non-sterile
feature
barrier: no
packaging
pack of 1000 ea (Bag)
manufacturer/tradename
Finn 9400230
maximum volume
250 μL
volume range
0.5-250 μL
Other Notes
Browse all pipette tips
Legal Information
Finnpipette is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Compare Similar Items
View Full Comparison
Show Differences
1 of 1
This Item
P2674P2924P9560
feature
barrier: no
feature
barrier: no
feature
barrier: no
feature
barrier: no
sterility
non-sterile
sterility
non-sterile
sterility
non-sterile
sterility
non-sterile
manufacturer/tradename
Finn 9400230
manufacturer/tradename
Finn 9401030
manufacturer/tradename
Finn 9402030
manufacturer/tradename
Finn 9401260
material
colorless
material
blue
material
colorless
material
orange
packaging
pack of 1000 ea (Bag)
packaging
pack of 1000 ea (Bag)
packaging
pack of 500 ea (Bag)
packaging
pack of 1000 ea (Bag)
volume range
0.5-250 μL
volume range
100-1000 μL
volume range
1-5 mL
volume range
5-300 μL
Still not finding the right product?
Explore all of our products under Finnpipette® pipette tips
Choose from one of the most recent versions:
Certificates of Analysis (COA)