All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C9H12N2O · HCl
CAS Number:
Molecular Weight:
200.67
Beilstein:
3915662
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Recommended Products
SMILES string
Cl.N[C@@H](Cc1ccccc1)C(N)=O
InChI
1S/C9H12N2O.ClH/c10-8(9(11)12)6-7-4-2-1-3-5-7;/h1-5,8H,6,10H2,(H2,11,12);1H/t8-;/m0./s1
InChI key
KLHLGTPNBQXSJT-QRPNPIFTSA-N
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
L-Phenylalaninamide hydrochloride is an amino acid amide.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
A V Chetkauskaĭte et al.
Zhurnal evoliutsionnoi biokhimii i fiziologii, 24(4), 465-470 (1988-07-01)
Synthesis of peptides during polymerization of GlyNH2 and PheNH2 has been demonstrated by means of gel-chromatography and thin-layer chromatography. The optima of pH and temperature have been estimated for the reaction. Grem's salt, tripolyphosphate and pyrophosphate were shown to cause
Seiji Okazaki et al.
Acta crystallographica. Section D, Biological crystallography, 64(Pt 3), 331-334 (2008-03-08)
The crystal structures of D-amino-acid amidase (DAA) from Ochrobactrum anthropi SV3 in complex with L-phenylalanine and with L-phenylalanine amide were determined at 2.3 and 2.2 A resolution, respectively. Comparison of the L-phenylalanine amide complex with the D-phenylalanine complex reveals that
Ravi Bhushan et al.
Biomedical chromatography : BMC, 23(12), 1291-1299 (2009-06-03)
Twelve chiral derivatizing reagents (CDRs) were synthesized by substituting one of the fluorine atoms in 1,5-difluoro-2,4-dinitrobenzene (DFDNB) with three optically pure amines [(R)-(-)-1-cyclohexylethylamine, (+)-dehydroabietylamine and (S)-(-)-alpha,4-dimethylbenzylamine], six amino acid amides [L-Ala-NH(2), L-Phe-NH(2), L-Val-NH(2), L-Leu-NH(2), L-Met-NH(2) and D-Phg-NH(2)] and three amino
A routine method for alkaline phosphatase assay in pregnancy serum.
D R Pledger et al.
Clinica chimica acta; international journal of clinical chemistry, 122(1), 71-74 (1982-06-16)
Z Chen et al.
Analytical chemistry, 73(14), 3348-3357 (2001-07-31)
A new type of chiral monolithic column was successfully developed for the enantioseparation of dansyl amino acids by ligand exchange-capillary electrochromatography (LE-CEC) in this work. The monolithic column matrix was prepared by a sol-gel process and then chemically modified with
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service