Skip to Content
Merck
All Photos(1)

Key Documents

15380

Sigma-Aldrich

Boc-Ala-OH

≥99.0% (TLC), for peptide synthesis

Synonym(s):

N-(tert-Butoxycarbonyl)-L-alanine, Boc-L-alanine

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€31.30
25 G
€97.90
100 G
€146.00

€31.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€31.30
25 G
€97.90
100 G
€146.00

About This Item

Linear Formula:
CH3CH[NHCO2C(CH3)3]CO2H
CAS Number:
Molecular Weight:
189.21
Beilstein:
1726365
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

€31.30


Check Cart for Availability

Request a Bulk Order

Product Name

Boc-Ala-OH, ≥99.0% (TLC)

Assay

≥99.0% (TLC)

optical activity

[α]20/D −25±1°, c = 2% in acetic acid

reaction suitability

reaction type: Boc solid-phase peptide synthesis
reaction type: C-H Activation
reagent type: ligand
reaction type: Peptide Synthesis

mp

79-83 °C (lit.)

application(s)

peptide synthesis

functional group

amine
carboxylic acid

SMILES string

C[C@H](NC(=O)OC(C)(C)C)C(O)=O

InChI

1S/C8H15NO4/c1-5(6(10)11)9-7(12)13-8(2,3)4/h5H,1-4H3,(H,9,12)(H,10,11)/t5-/m0/s1

InChI key

QVHJQCGUWFKTSE-YFKPBYRVSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Boc-Ala-OH can be used:
  • In the preparation of N-propargylalanine, a key precursor to generate N-(3-aryl)propylated alanine residues.[1]
  • In the resolution of racemic mixture of 3,3′-bis(benzyloxy)-1,1′-binaphthalene-2,2′-diol.[2]
  • In the one-pot synthesis of hybrid tripeptidomimetics containing both amide and imide functionalities.[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of N-(3-arylpropyl) amino acid derivatives by sonogashira types of reaction in aqueous media
Lopez-Deber MP, et al.
Organic Letters, 3(18), 2823-2826 (2001)
Convenient synthesis and efficient resolution of 3, 3′-bis (benzyloxy)-1, 1′-binaphthalene-2, 2′-diol
Tsubaki K, et al.
Tetrahedron Asymmetry, 14(10), 1393-1396 (2003)
A facile one pot route for the synthesis of imide tethered peptidomimetics
Panduranga V, et al.
Organic & Biomolecular Chemistry, 14(2), 556-563 (2016)
Leila Quercini et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 34(1), 192-207 (2020-01-10)
The peptide sequence KKIRVRLSA was synthesized in a dimeric structure (SET-M33DIM) and evaluated as a candidate drug for infections due to multidrug-resistant (MDR) Gram-negative pathogens. SET-M33DIM showed significant antibacterial activity against MDR strains of Klebsiella pneumoniae, Acinetobacter baumannii, and Escherichia
Huang-Han Chen et al.
Analytica chimica acta, 865, 53-59 (2015-03-04)
Polydimethylsiloxane (PDMS) is widely used for microfabrication and bioanalysis; however, its surface functionalization is limited due to the lack of active functional groups and incompatibility with many solvents. We presented a novel approach for in situ fabrication of cleavable peptide

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service