Skip to Content
Merck
All Photos(1)

Key Documents

P222

Sigma-Aldrich

Pandinotoxin-Kα

>99%, powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C169H267N53O48S7
CAS Number:
Molecular Weight:
4033.71
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

Assay

>99%

form

powder

color

off-white

storage temp.

−20°C

Amino Acid Sequence

Thr-Ile-Ser-Cys-Thr-Asn-Pro-Lys-Gln-Cys-Tyr-Pro-His-Cys-Lys-Lys-αGlu-Thr-Gly-Tyr-Pro-Asn-Ala-Lys-Cys-Met-Asn-Arg-Lys-Cys-Lys-Cys-Phe-Gly-Arg

General description

Pandinotoxin (PiTX)-Kα is a 35 amino acid peptide isolated from venom of the scorpion Pandinus imperaton. PiTX-Kβ and PiTXγ are other two members of pandinotoxin group. All the three pandinotoxins have structures similar to another scorpion toxin charybdotoxin (ChTX). The structure of PiTXs contains a β-sheet at the C- terminal end with three cysteines which is linked to the central α-helix by two disulfide bridges (C17-C35 and C13-C33) and to an extended amino-terminal fragment by the third disulfide bridge (C7-C28).

Biochem/physiol Actions

Pandinotoxin (PiTX) –Kα along with PiTX-Kβ and PiTXγ plays a vital role in blocking voltage-gated K+ channels.

Legal Information

Sold with the permission of University of Maryland.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D R Matteson et al.
The Journal of physiology, 503 ( Pt 2), 285-301 (1997-11-05)
1. The ability of three structurally homologous scorpion toxins to block voltage-dependent K+ currents in rat dorsal root ganglion neurones was examined using the patch-clamp technique. 2. Neurones with a diameter > 35 microns had two identifiable components of macroscopic
R S Rogowski et al.
Molecular pharmacology, 50(5), 1167-1177 (1996-11-01)
Three 35-amino acid peptide K+ channel toxins (pandinotoxins) were purified from the venom of the scorpion Pandinus imperaton the toxins are designated pandinotoxin (PiTX)-K alpha, PiTX-K beta, and PiTX-K gamma. In an 86Rb tracer flux assay on rat brain synaptosomes
T C Tenenholz et al.
Biochemistry, 36(10), 2763-2771 (1997-03-11)
PiTX-K alpha, a 35-residue peptide recently isolated from the venom of Pandinus imperator, blocks the rapidly inactivating (A-type) K+ channel(s) in rat brain synaptosomes and the cloned Kv 1.2 potassium channel at very low toxin concentrations (6 nM and 32

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service