Skip to Content
Merck
All Photos(3)

Key Documents

P2222

Sigma-Aldrich

Potassium phosphate dibasic

meets USP testing specifications

Synonym(s):

Dipotassium hydrogenphosphate, Dipotassium phosphate, sec.-Potassium phosphate

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€86.70
500 G
€269.00
1 KG
€499.00

€86.70


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
€86.70
500 G
€269.00
1 KG
€499.00

About This Item

Linear Formula:
K2HPO4
CAS Number:
Molecular Weight:
174.18
EC Number:
MDL number:
UNSPSC Code:
12161700
eCl@ss:
38070212
PubChem Substance ID:
NACRES:
NA.25

€86.70


Check Cart for Availability

Request a Bulk Order

Agency

USP/NF
meets USP testing specifications

Quality Level

Assay

98.0-100.5%

form

granular

application(s)

pharmaceutical (small molecule)

SMILES string

[K+].[K+].OP([O-])([O-])=O

InChI

1S/2K.H3O4P/c;;1-5(2,3)4/h;;(H3,1,2,3,4)/q2*+1;/p-2

InChI key

ZPWVASYFFYYZEW-UHFFFAOYSA-L

Application


  • Chitosan-PVA-PVP/nano-clay composite: a promising tool for controlled drug delivery.: This study explores the development of a composite material using chitosan, PVA, PVP, and potassium phosphate dibasic for controlled drug delivery. The composite demonstrates enhanced drug encapsulation and release properties, making it suitable for pharmaceutical applications (Ali et al., 2024).

  • Biocatalysis with in-situ product removal improves p-coumaric acid production.: This research investigates the use of potassium phosphate dibasic in biocatalytic processes for the production of p-coumaric acid. The study highlights the efficiency of in-situ product removal techniques to enhance production yields in biochemistry applications (Virklund et al., 2024).

  • Partitioning recombinant chitinase from Nicotiana benthamiana by an aqueous two-phase system based on polyethylene glycol and phosphate salts.: This research employs potassium phosphate dibasic in an aqueous two-phase system for the extraction and purification of recombinant chitinase. The study demonstrates the effectiveness of this method in biotechnological applications (Tue et al., 2024).

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ming Ye et al.
Biosensors & bioelectronics, 73, 208-213 (2015-06-17)
Several volatile organic compounds (VOCs) are released from human breath or skin. Like chemical substances in blood or urine, some of these vapors can provide valuable information regarding the state of the human body. A highly sensitive acetone biochemical gas
Pamela Gamba et al.
PLoS genetics, 11(6), e1005047-e1005047 (2015-06-26)
Gene expression can be highly heterogeneous in isogenic cell populations. An extreme type of heterogeneity is the so-called bistable or bimodal expression, whereby a cell can differentiate into two alternative expression states. Stochastic fluctuations of protein levels, also referred to
Kenshi Hayakawa et al.
Journal of bioscience and bioengineering, 120(5), 532-538 (2015-04-29)
S-Adenosyl-L-methionine (SAM) is a major biological methyl group donor, and is used as a nutritional supplement and prescription drug. Yeast is used for the industrial production of SAM owing to its high intracellular SAM concentrations. To determine the regulation mechanisms
Audrey Beaussart et al.
ACS nano, 8(10), 10723-10733 (2014-10-07)
A variety of bacterial pathogens use nanoscale protein fibers called type IV pili to mediate cell adhesion, a primary step leading to infection. Currently, how these nanofibers respond to mechanical stimuli and how this response is used to control adhesion
Ragnar Thomsen et al.
Drug testing and analysis, 7(7), 565-576 (2014-10-28)
Synthetic cannabimimetic agents are a large group of diverse compounds which act as agonists at cannabinoid receptors. Since 2004, synthetic cannabinoids have been used recreationally, although several of the compounds have been shown to cause severe toxicity in humans. In

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service