Skip to Content
Merck
All Photos(1)

Documents

P0549

Sigma-Aldrich

Microcapillary tube, calibrated

capacity 1-5 μL

Synonym(s):

Microcapillary Tube

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500
NACRES:
NB.01

material

glass

packaging

pack of 1
pack of 250 ea

manufacturer/tradename

Drummond Scientific 2-000-001

measuring range

accuracy: ±0.5%

capacity

1-5 μL

color

color-coded white

Looking for similar products? Visit Product Comparison Guide

General description

  • Drummond Calibrated Micropipettes provide accuracy and convenience for a wide variety of liquid transfer applications.
  • Made of high quality glass.
  • Fire polished micropipettes are imprinted with volume and a color-code bar for easy identification.
  • Calibrated "To Deliver."

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Steffy B Manjila et al.
Current biology : CB, 29(2), 283-293 (2019-01-08)
Insect flight is a complex behavior that requires the integration of multiple sensory inputs with flight motor output. Although previous genetic studies identified central brain monoaminergic neurons that modulate Drosophila flight, neuro-modulatory circuits underlying sustained flight bouts remain unexplored. Certain
Naveen Prasad et al.
Cell reports, 24(12), 3194-3206 (2018-09-21)
A balanced diet of macronutrients is critical for animal health. A lack of specific elements can have profound effects on behavior, reproduction, and lifespan. Here, we used Drosophila to understand how the brain responds to carbohydrate deprivation. We found that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service