Skip to Content
Merck
All Photos(1)

Key Documents

Z611239

Hirschmann® microcapillary pipette

tube capacity 1-5 μL

Synonym(s):

Capillary pipette, Hirschmann pipette, Microcapillary pipette

Sign Into View Organizational & Contract Pricing

Select a Size

250 EA
€34.40

€34.40


Check Cart for Availability


Select a Size

Change View
250 EA
€34.40

About This Item

UNSPSC Code:
41121500
NACRES:
NB.01

€34.40


Check Cart for Availability

material

glass
white

description

Genuine Schott® Duran glass

packaging

pack of 250 ea

manufacturer/tradename

Hirschmann 9600105

availability

not available in USA and Canada

technique(s)

thin layer chromatography (TLC): suitable

L

125 mm

tube capacity

1-5 μL

Looking for similar products? Visit Product Comparison Guide

General description

Disposable pipettes with ringmark. The exact ringmark guarantees an accurate volume determination. Each micro capillary is color-coded to avoid mix ups. Useful for spotting TLC plates.

Legal Information

Hirschmann is a registered trademark of Hirschmann Laborgeräte GmbH & Co.
Schott is a registered trademark of Schott AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asier Aristieta et al.
Current biology : CB, 31(4), 707-721 (2020-12-12)
The basal ganglia (BG) inhibit movements through two independent circuits: the striatal neuron-indirect and the subthalamic nucleus-hyperdirect pathways. These pathways exert opposite effects onto external globus pallidus (GPe) neurons, whose functional importance as a relay has changed drastically with the
Brice de la Crompe et al.
Nature communications, 11(1), 1570-1570 (2020-03-29)
The dynamical properties of cortico-basal ganglia (CBG) circuits are dramatically altered following the loss of dopamine in Parkinson's disease (PD). The neural circuit dysfunctions associated with PD include spike-rate alteration concomitant with excessive oscillatory spike-synchronization in the beta frequency range

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service