Skip to Content
Merck
All Photos(1)

Documents

P0157

Sigma-Aldrich

Phosphatase, Acid from potato

ammonium sulfate suspension, ≥200 units/mg protein (biuret)

Synonym(s):

(Orthophosphoric-monoester phosphohydrolase acid optimum), Acid Phosphatase from potato

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204

form

ammonium sulfate suspension

specific activity

≥200 units/mg protein (biuret)

mol wt

111 kDa

foreign activity

Apyrase (ATPase) ≤5% of base activity

storage temp.

2-8°C

InChI

1S/C6H10O2/c1-3-4-8-5-6(2)7/h1,6-7H,4-5H2,2H3

InChI key

GZCWLCBFPRFLKL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acid phosphatase from potatoes is a 111 kDa diner consisting of two subunits at 41 and 35 kDa. This phosphatase has also been shown to cleave DNA.

Application

Phosphatase acid from potato has been used in a study to develop a method of efficient enzymatic hydrolysis of polyprenyl pyrophosphates. It has also been used in a study to investigate the kinetics of the hydrolysis of sodium p-nitrophenylphosphate and other phosphoric acid monoesters.

Biochem/physiol Actions

Acid phosphatases (APase) are a family of enzymes that non-specifically catalyze the hydrolysis of monoesters and anhydrides of phosphoric acid to produce inorganic phosphate at an optimum pH of 4 to 7.

Unit Definition

One unit will hydrolyze 1.0 μmole of p-nitrophenyl phosphate per min at pH 4.8 at 37 °C.

Physical form

Suspension in 1.8 M (NH4)2SO4, 10 mM MgCl2, pH 5.5

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Insight into bio-physiological functions of acid phosphatase from potatoes (Solanum tuberosum): A bioanalytical approach.
Rane, S.
Indian Journal of Chemistry, 48a, 15-24 (2009)
Y Mikuni-Takagaki et al.
The Biochemical journal, 268(3), 585-591 (1990-06-15)
In order to understand the mechanism of the post-translational processing of bone phosphoproteins in embryonic bone, periosteal bone strips isolated from 12-day-embryonic-chick tibiae were cultured and the bone proteins labelled with Na2H32PO4. Of the total radiolabelled proteins recovered from the
H Fujii et al.
Biochimica et biophysica acta, 712(3), 716-718 (1982-09-14)
A method of efficient enzymatic hydrolysis of polyprenyl pyrophosphates was established. Polyprenyl pyrophosphates such as all-trans-octaprenyl and all-trans-nonaprenyl (solanesyl) pyrophosphate can be hydrolyzed completely to the corresponding primary alcohols by the action of potato acid phosphatase in the presence of
The kinetics and mechanism of the hydrolysis of phosphoric acid esters by potato acid phosphatase.
E F ALVAREZ
Biochimica et biophysica acta, 59, 663-672 (1962-06-04)
Y Sugiura et al.
The Journal of biological chemistry, 256(20), 10664-10670 (1981-10-25)
A new manganese-containing acid phosphatase has been isolated and crystallized from sweet potato tubers. The pure enzyme contains one atom of manganese per Mr = 110,000 polypeptide and shows phosphatase activity toward various phosphate substrates. The pH optimum of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service