Skip to Content
Merck
All Photos(1)

Key Documents

524529

Sigma-Aldrich

Phosphatase, Acid, Potato

Synonym(s):

Phosphatase, Acid, Potato

Sign Into View Organizational & Contract Pricing

Select a Size

1 KU
€579.00

€579.00


Estimated to ship on24 May 2025



Select a Size

Change View
1 KU
€579.00

About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.54

€579.00


Estimated to ship on24 May 2025


form

lyophilized

Quality Level

specific activity

≥2 U/mg

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

solubility

3.2 M ammonium sulfate, pH 6.0, containing 1% BSA: soluble
6% BSA in water: soluble

shipped in

ambient

storage temp.

2-8°C

SMILES string

O(CC(O)C)CC#C

InChI

1S/C6H10O2/c1-3-4-8-5-6(2)7/h1,6-7H,4-5H2,2H3

InChI key

GZCWLCBFPRFLKL-UHFFFAOYSA-N

General description

Native potato acid phosphatase. May be used for the removal of phosphate groups from phosphoproteins, such as β-casein, ovalbumin, and pepsinogen.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will hydrolyze 1 µmol of pNPP per min at 25°C, pH 5.6.

Reconstitution

Following reconstitution, refrigerate (4°C). Stock solutions in ammonium sulfate are stable for up to 4 months at 4°C. Stock solutions in aqueous BSA should be used immediately upon reconstitution.

Other Notes

Bingham, E.W., et al. 1976. Biochim. Biophys. Acta429, 448.
Bessey, O.A. 1946. J. Biol. Chem.164, 32.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service