Skip to Content
Merck

Skip To

P0037

PP242 hydrate

≥98% (HPLC), powder

Synonym(s):

2-(4-amino-1-isopropyl-1H-pyrazolo[3,4-d]pyrimidin-3-yl)-1H-indol-5-ol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Check Cart for Availability
€ 244,00

About This Item

Empirical Formula (Hill Notation):
C16H16N6O · xH2O
CAS Number:
Molecular Weight:
308.34 (anhydrous basis)
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥98% (HPLC)
Form:
powder

€ 244,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

off-white

solubility

DMSO: >10 mg/mL

storage temp.

2-8°C

SMILES string

O.CC(C)n1nc(-c2cc3cc(O)ccc3[nH]2)c4c(N)ncnc14

InChI

1S/C16H16N6O.H2O/c1-8(2)22-16-13(15(17)18-7-19-16)14(21-22)12-6-9-5-10(23)3-4-11(9)20-12;/h3-8,20,23H,1-2H3,(H2,17,18,19);1H2

InChI key

FMDOLJSFPXFBIL-UHFFFAOYSA-N

Application

PP242 hydrate has been used:
  • as a mammalian target of rapamycin (mTOR) inhibitor to study its effects on rat fibroblast mTOR complex 1 and 2 (mTORC1 & 2) signaling
  • as an mTOR inhibitor to treat mouse embryonic fibroblasts (MEF) and cumulus cells (CCs) to study its effects on reactivation of embryonic nucleoli and ribosome biogenesis
  • as a target of rapamycin (TOR) kinase inhibitor for the treatment of Arabidopsis to study its effects against Fusarium graminearum infection

Biochem/physiol Actions

PP242 is a potent and selective mTOR inhibitor.
PP242 is a potent and selective mTOR inhibitor. PP242 is a first selective inhibitor that targets ATP domain of mTOR.

Features and Benefits

This compound is featured on the PKB/Akt page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P0042P0036SML1381
assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

solubility

DMSO: >10 mg/mL

solubility

DMSO: >10 mg/mL

solubility

DMSO: >10 mg/mL

solubility

H2O: 10 mg/mL, clear, DMSO: 5 mg/mL, clear (warmed)

color

off-white

color

white to off-white

color

off-white

color

white to beige


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. I would like to get information about the water content of the compound. It is written as X in the compound formula. How many water molecules are found in the PP242 hydrate molecule?

    1 answer
    1. The water content can be found on the lot-dependent Certificate of Analysis.

      Helpful?

  2. What is the degree of hydration for this product?

    1 answer
    1. The specification for the degree of hydration for this product is an elemental analysis value of H2O (moles) ranging from 0.0 to 3.0. The actual value can be found in the batch-specific Certificate of Analysis.

      Helpful?

Reviews

No rating value

Active Filters