Skip to Content
Merck
All Photos(1)

Key Documents

N8914

Sigma-Aldrich

NSC308848

≥98% (HPLC), powder

Synonym(s):

5-amino-2-(2-(diethylamino)ethyl)-1H-benzo[de]isoquinoline-1,3(2H)-dione

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€130.00

€130.00


Check Cart for Availability


Select a Size

Change View
5 MG
€130.00

About This Item

Empirical Formula (Hill Notation):
C18H21N3O2
CAS Number:
Molecular Weight:
311.38
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€130.00


Check Cart for Availability

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

yellow

solubility

DMSO: >20 mg/mL

storage temp.

2-8°C

SMILES string

CCN(CC)CCN1C(=O)c2cccc3cc(N)cc(C1=O)c23

InChI

1S/C18H21N3O2/c1-3-20(4-2)8-9-21-17(22)14-7-5-6-12-10-13(19)11-15(16(12)14)18(21)23/h5-7,10-11H,3-4,8-9,19H2,1-2H3

InChI key

QDFFNSUDGUZZSJ-UHFFFAOYSA-N

Biochem/physiol Actions

NSC308848 is a small molecular transcription factor inhibitor.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service