Skip to Content
Merck
All Photos(1)

Key Documents

235598

Sigma-Aldrich

4-Amino-1,8-naphthalic anhydride

95%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€237.00

€237.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€237.00

About This Item

Empirical Formula (Hill Notation):
C12H7NO3
CAS Number:
Molecular Weight:
213.19
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€237.00


Check Cart for Availability

Request a Bulk Order

Assay

95%

form

powder

impurities

1-2% DMF

SMILES string

Nc1ccc2C(=O)OC(=O)c3cccc1c23

InChI

1S/C12H7NO3/c13-9-5-4-8-10-6(9)2-1-3-7(10)11(14)16-12(8)15/h1-5H,13H2

InChI key

QIXHMCMCFSNKOG-UHFFFAOYSA-N

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service