Skip to Content
Merck
All Photos(2)

Documents

N8784

Sigma-Aldrich

Norcantharidin

solid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8O4
CAS Number:
Molecular Weight:
168.15
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

form

solid

SMILES string

O=C1OC(=O)C2C3CCC(O3)C12

InChI

1S/C8H8O4/c9-7-5-3-1-2-4(11-3)6(5)8(10)12-7/h3-6H,1-2H2

InChI key

JAABVEXCGCXWRR-UHFFFAOYSA-N

Gene Information

human ... PPP2R5A(5525)

General description

Norcantharidin (NCTD) is a demethylated derivative of cantharidin (CTD).

Application

Norcantharidin has been used in primary cell culture and treatment. It has also been used to treat mice inoculated with Huh7 cells subcutaneously.

Biochem/physiol Actions

Norcantharidin is a potential antitumor agent. Norcantharidin helps to block SK-N-SH neuroblastoma cell growth by stimulating autophagy and apoptosis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yong-yan Bei et al.
International journal of nanomedicine, 7, 1819-1827 (2012-05-24)
In this paper, two novel liver-targeting nanoparticles, norcantharidin-loaded chitosan nanoparticles (NCTD-CS-NPs) and norcantharidin-associated galactosylated chitosan nanoparticles (NCTD-GC-NPs), were prepared using ionic cross-linkage. The physical properties, particle size, encapsulation efficiency, and drug release characteristics of the nanoparticles were investigated in vitro.
Xin-Yuan Ding et al.
International journal of nanomedicine, 7, 1723-1735 (2012-05-24)
A novel formulation containing polyvinylpyrrolidone (PVP) K(30)-coated norcantharidin (NCTD) chitosan nanoparticles (PVP-NCTD-NPs) was prepared by ionic gelation between chitosan and sodium tripolyphosphate. The average particle size of the PVP-NCTD-NPs produced was 140.03 ± 6.23 nm; entrapment efficiency was 56.33% ±
Mark Tarleton et al.
European journal of medicinal chemistry, 54, 573-581 (2012-07-17)
Cantharidin (1) and norcantharidin (2) display high levels of anticancer activity against a broad range of tumour cell lines. Synthetic manipulation of norcantharidin yields (3S,3aR,4S,7R,7aS)-3-hydroxyhexahydro-4,7-epoxyisobenzofuran-1(3H)-one (3), which also displays a high level of anticancer activity against tumour cells but interestingly
Yong Li et al.
Frontiers in pharmacology, 11, 187-187 (2020-03-21)
Drug repositioning, development of new uses for marketed drugs, is an effective way to discover new antitumor compounds. In this study, we used a new method, filtering compounds via molecular docking to find key targets combination. The data of gene
Min Guan et al.
International journal of nanomedicine, 7, 1921-1930 (2012-05-19)
In this paper, novel liver-targeting nanoparticles (NPs), lactosyl-norcantharidin (Lac-NCTD)-associated N-trimethyl chitosan (TMC) NPs (Lac-NCTD-TMC-NPs), were prepared using ionic cross-linkage. The physical properties, particle size, and encapsulation efficiency of the nanoparticles were then investigated. The continuous line of heterogeneous human epithelial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service