Skip to Content
Merck
All Photos(1)

Key Documents

8.22002

Sigma-Aldrich

Crotonic anhydride

(mixture of cis and trans isomers) for synthesis

Synonym(s):

Crotonic anhydride, 2-Butenoic anhydride

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€37.80

€37.80


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
€37.80

About This Item

Linear Formula:
(CH3CH=CHCO)2O
CAS Number:
Molecular Weight:
154.16
MDL number:
UNSPSC Code:
12352106
EC Index Number:
210-807-1
NACRES:
NA.22

€37.80


Estimated to ship on29 May 2025


Request a Bulk Order

vapor pressure

1 hPa ( 25 °C)

Quality Level

form

liquid

autoignition temp.

290 °C

potency

2061 mg/kg LD50, oral (Rat)

bp

250 °C

mp

-20 °C

transition temp

flash point 113 °C

density

1.04 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(C(=O)\C=C\C)C(=O)\C=C\C

InChI

1S/C8H10O3/c1-3-5-7(9)11-8(10)6-4-2/h3-6H,1-2H3/b5-3+,6-4+

InChI key

VJDDQSBNUHLBTD-GGWOSOGESA-N

General description

Crotonic anhydride is also known as 2-butenoic anhydride is commonly used as a precursor in the synthesis of polymers. Its reactive anhydride group makes it an effective acylating agent in esterification reactions, facilitating the formation of esters by reacting with alcohols.

Analysis Note

Assay isomers (GC, area%): ≥ 95.5 % (a/a)
Density (d 20 °C/ 4 °C): 1.038 - 1.040
Identity (IR): passes test

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F

Flash Point(C)

113 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service