Skip to Content
Merck
All Photos(1)

Key Documents

N7032

Sigma-Aldrich

NFAT Inhibitor

>96% (HPLC), solid

Synonym(s):

VIVIT

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€531.00

€531.00


Estimated to ship on20 May 2025



Select a Size

Change View
1 MG
€531.00

About This Item

Empirical Formula (Hill Notation):
C75H118N20O22S
CAS Number:
Molecular Weight:
1683.93
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.26

€531.00


Estimated to ship on20 May 2025


Quality Level

Assay

>96% (HPLC)

form

solid

storage temp.

−20°C

SMILES string

S(CC[C@H](N)C(=O)N[C@@H](C)C(=O)NCC(=O)N1[C@@H](CCC1)C(=O)N[C@@H](Cc5nc[nH]c5)C(=O)N2[C@@H](CCC2)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H]([C@H](CC)C)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H]([C@H](CC)C)C(=O)N[C@@H]([C@H](O)C)C(=O)NCC(=O)N3[C@@H](CCC3)C(=O)N[C@@H](Cc4nc[nH]c

InChI

1S/C75H118N20O22S/c1-12-39(7)59(90-70(111)57(37(3)4)88-68(109)52-19-16-27-95(52)74(115)49(30-44-32-78-36-82-44)87-67(108)51-18-15-26-94(51)53(97)33-79-62(103)41(9)83-63(104)45(76)24-28-118-11)72(113)89-58(38(5)6)71(112)91-60(40(8)13-2)73(114)92-61(42(10)96)69(110)80-34-54(98)93-25-14-17-50(93)66(107)86-48(29-43-31-77-35-81-43)65(106)84-46(20-22-55(99)100)64(105)85-47(75(116)117)21-23-56(101)102/h31-32,35-42,45-52,57-61,96H,12-30,33-34,76H2,1-11H3,(H,77,81)(H,78,82)(H,79,103)(H,80,110)(H,83,104)(H,84,106)(H,85,105)(H,86,107)(H,87,108)(H,88,109)(H,89,113)(H,90,111)(H,91,112)(H,92,114)(H,99,100)(H,101,102)(H,116,117)/t39-,40-,41-,42+,45-,46-,47-,48-,49-,50-,51-,52-,57-,58-,59-,60-,61-/m0/s1

InChI key

QPMHUXBSHGAVGD-MCDIZDEASA-N

Gene Information

Amino Acid Sequence

Met-Ala-Gly-Pro-His-Pro-Val-Ile-Val-Ile-Thr-Gly-Pro-His-Glu-Glu

Application

NFAT Inhibitor, VIVIT has been used to study the signaling pathways involved in controlling inhibitory molecules and subsequent onset of T-cell impairment.[1]
NFAT inhibitor has been as an inhibitor of nuclear factor of activated T cells.[2]

Biochem/physiol Actions

High-affinity calcineurin-binding peptide that inhibits NFAT (Nuclear Factor of Activated T cells) activation and NFAT-dependent expression of endogenous cytokine genes in T cells.
Inhibits NFAT (Nuclear Factor of Activated T cells) activation and NFAT-dependent expression of endogenous cytokine genes in T cells.
Nuclear factor of activated T cells (NFAT) participates in the activity and growth of immune and non-immune cells.[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

p38 Mitogen-Activated Protein Kinase/Signal Transducer and Activator of Transcription-3 Pathway Signaling Regulates Expression of Inhibitory Molecules in T Cells Activated by HIV-1?Exposed Dendritic Cells
Karlhans Fu Che
Molecular Medicine (2012)
Carmela Gallo et al.
Cellular and molecular life sciences : CMLS, 79(7), 369-369 (2022-06-21)
The immune response arises from a fine balance of mechanisms that provide for surveillance, tolerance, and elimination of dangers. Sulfavant A (SULF A) is a sulfolipid with a promising adjuvant activity. Here we studied the mechanism of action of SULF
Adolf E Schindler
International journal of women's health, 3, 175-184 (2011-07-28)
Endometriosis is a chronic disease primarily affecting women of childbearing age, in which endometriotic lesions form outside the uterus, typically leading to painful symptoms, fatigue, and infertility. The symptoms of endometriosis may cause significant impairment in quality of life and
Karlhans Fru Che et al.
Molecular medicine (Cambridge, Mass.), 18, 1169-1182 (2012-07-11)
Human immunodeficiency virus type 1 (HIV-1) infection enhances the expression of inhibitory molecules on T cells, leading to T-cell impairment. The signaling pathways underlying the regulation of inhibitory molecules and subsequent onset of T-cell impairment remain elusive. We showed that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service