Skip to Content
Merck
All Photos(1)

Key Documents

480401-M

Sigma-Aldrich

NFAT Inhibitor, Cell-Permeable

The NFAT Inhibitor, Cell-Permeable controls the biological activity of NFAT. This small molecule/inhibitor is primarily used for Inflammation/Immunology applications.

Synonym(s):

NFAT Inhibitor, Cell-Permeable, 11R-VIVIT, Nuclear Factor of Activated T cells Inhibitor, RRRRRRRRRRR-GGG- MAGPHPVIVITGPHEE, 11R-VIVIT, Nuclear Factor of Activated T cells Inhibitor, RRRRRRRRRRR-GGG-MAGPHPVIVITGPHEE

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€484.00

€484.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
€484.00

About This Item

Empirical Formula (Hill Notation):
C147H259N67O36S
Molecular Weight:
3573.12
UNSPSC Code:
12352202
NACRES:
NA.77

€484.00


Estimated to ship on25 May 2025


Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

water: 1 mg/mL
DMSO: 5 mg/mL

storage temp.

−20°C

General description

A cell-permeable version of NFAT Inhibitor (Cat. No. 480402) that is modified at the N-terminal with an eleven arginine transduction domain and a three glycine linker sequence. Prevents the activation and proliferation of T cells both in vitro ( ~ 43% at 1 µM using mixed lymphocyte cultures) and in vivo (10 mg/kg using C3H/HeN mice). Shown to be effective in providing immunosuppression in a murine islet xenograft model. Unlike Cyclosporin A (Cat. No. 239835) and FK506, 11R-VIVIT does not cause side effects outside the immune system as a result of calcineurin (Cat. No. 539568) inhibition and therefore serves as a less toxic alternative for in vivo applications. The functional half-life of 11R-VIVIT has been estimated to be ~30 h in Jurkat cells.

Application

Use at 1 µM for preventing the activation and proliferation of T cells

Biochem/physiol Actions

Primary Target
NFAT

Warning

Toxicity: Harmful (C)

Sequence

H-Arg-Arg-Arg-Arg-Arg-Arg-Arg-Arg-Arg-Arg-Arg-Gly-Gly-Gly-Met-Ala-Gly-Pro-His-Pro-Val-Ile-Val-Ile-Thr-Gly-Pro-His-Glu-Glu-OH

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Noguchi, H., et al. 2004. Nat. Med.10, 305.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service