Skip to Content
Merck
All Photos(1)

Documents

N5386

Sigma-Aldrich

Nuclease micrococcal from Staphylococcus aureus

100-300 units/mg protein

Synonym(s):

Endonuclease micrococcal, MNase, Micrococcal endonuclease

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

Staphylococcus aureus

Quality Level

form

powder

specific activity

100-300 units/mg protein

composition

Protein, ≥40% E1%/280 (Balance primarily sodium acetate)

technique(s)

DNA extraction: suitable
DNA purification: suitable

suitability

suitable for molecular biology

application(s)

cell analysis

storage temp.

−20°C

Gene Information

Staphylococcus aureus subsp. aureus Mu50 ... nuc(1120790)

Looking for similar products? Visit Product Comparison Guide

Application

Micrococcal Nuclease can be used for the digestion of chromatin to identify the location of nucleosomes.
Nuclease micrococcal from Staphylococcus aureus has been used in a study to monitor hybridization reactions with DNA. It has also been used in a study to investigate the optimal conditions for assay of staphylococcal nuclease.

Biochem/physiol Actions

Hydrolyzes 5′-phosphodiester bonds of DNA.

Other Notes

Note: One μmolar unit = approx. 85 A260 units, where an A260 unit is equivalent to a ΔA260 of 1.0 in 30 min at pH 8.8 at 37 °C in a 3.55 mL reaction volume. Light path = 1 cm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Use of micrococcal nuclease to monitor hybridization reactions with DNA.
D L Kacian et al.
Analytical biochemistry, 58(2), 534-540 (1974-04-01)
Optimal conditions for assay of staphylococcal nuclease
KAMMAN, J. and S. TATINI
Journal of Food Science, 42, 421-424 (1977)
Ho-Ryun Chung et al.
PloS one, 5(12), e15754-e15754 (2011-01-06)
Eukaryotic genomes are packed into chromatin, whose basic repeating unit is the nucleosome. Nucleosome positioning is a widely researched area. A common experimental procedure to determine nucleosome positions involves the use of micrococcal nuclease (MNase). Here, we show that the
Luye Qin et al.
Nature communications, 12(1), 6589-6589 (2021-11-17)
ASH1L, a histone methyltransferase, is identified as a top-ranking risk factor for autism spectrum disorder (ASD), however, little is known about the biological mechanisms underlying the link of ASH1L haploinsufficiency to ASD. Here we show that ASH1L expression and H3K4me3
Mande Xue et al.
Molecular plant, 14(11), 1799-1813 (2021-07-10)
Global warming poses a major threat to plant growth and crop production. In some plants, including Arabidopsis thaliana, elevated temperatures induce a series of morphological and developmental adjustments termed thermomorphogenesis, which facilitates plant cooling under high-temperature conditions. Plant thermal response is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service