Skip to Content
Merck
All Photos(1)

Key Documents

N3790

Sigma-Aldrich

Nutrient Mixture F-12 Ham

StableCell, with stable glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

Nutrient Mixture F-12 Ham, Ham’s F-12

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
€44.20

€44.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 ML
€44.20

About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

€44.20


Check Cart for Availability

Request a Bulk Order

Product Name

StableCell Ham′s F-12, With stable glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

Endotoxin, tested

components

HEPES: no
sodium pyruvate: 0.11 g/L
stable glutamine: yes
phenol red: yes
NaHCO3: yes

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Developed for growth of primary rat hepatocytes and rat prostate epithelial cells. Also used in a clonal toxicity assay using CHO cells.
This Nutrient Mixture F-12 Hams differs from the original formula, because it is supplemented with L-alanyl-L-glutamine dipeptide. Glutamine is notoriously unstable in cell culture. This dipeptide provides a more stable form of glutamine for use in cell cultures.

Legal Information

StableCell is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quanchi Chen et al.
Cancers, 12(3) (2020-03-08)
The ruthenium-based photosensitizer (PS) TLD1433 has completed a phase I clinical trial for photodynamic therapy (PDT) treatment of bladder cancer. Here, we investigated a possible repurposing of this drug for treatment of conjunctival melanoma (CM). CM is a rare but
Tsuyoshi Mishiro et al.
The EMBO journal, 28(9), 1234-1245 (2009-03-27)
Long-range regulatory elements and higher-order chromatin structure coordinate the expression of multiple genes in cluster, and CTCF/cohesin-mediated chromatin insulator may be a key in this regulation. The human apolipoprotein (APO) A1/C3/A4/A5 gene region, whose alterations increase the risk of dyslipidemia
Fumitaka Kawakami et al.
PloS one, 7(1), e30834-e30834 (2012-02-04)
Leucine-rich repeat kinase 2 (LRRK2), a large protein kinase containing multi-functional domains, has been identified as the causal molecule for autosomal-dominant Parkinson's disease (PD). In the present study, we demonstrated for the first time that (i) LRRK2 interacts with tau
Henri J Huttunen et al.
Journal of molecular neuroscience : MN, 37(1), 6-15 (2008-07-12)
A common pathogenic event that occurs in all forms of Alzheimer's disease is the progressive accumulation of amyloid beta-peptide (Abeta) in brain regions responsible for higher cognitive functions. Inhibition of acyl-coenzyme A: cholesterol acyltransferase (ACAT), which generates intracellular cholesteryl esters

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service