Skip to Content
Merck
All Photos(1)

Key Documents

51448C

SAFC

Dulbecco′s Modified Eagle′s Medium/Ham′s Nutrient Mixture F12

suitable for cell culture

Pharma Manufacturing

Synonym(s):

Dulbecco’s Modified Eagle’s Medium/Nutrient Mixture F-12 Ham, DME/F-12, 1:1 mixture

Sign Into View Organizational & Contract Pricing

Select a Size

1000 ML
€50.00

€50.00


Check Cart for Availability


Select a Size

Change View
1000 ML
€50.00

About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

€50.00


Check Cart for Availability

description

for research or for further manufacturing use
with 3151 mg/L dextrose, with 2.5 mM L-glutamine, with 55 mg/L sodium pyruvate

Quality Level

sterility

sterile

form

liquid

technique(s)

cell culture | mammalian: suitable
cell culture | stem cell: suitable

components

phenol red: yes
L-glutamine: yes
sodium pyruvate: 0.055 g/L
glucose: 3.15 g/L

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

DMEM/F12 was originally developed to study hormonal requirements of cells in culture.

Application

Dulbecco′s Modified Eagle′s Medium/Ham′s Nutrient Mixture F12 has been used a primary culture media for propagation of adipose-derived mesenchymal stem cells.[1]

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The effect of metamizole and tolfenamic acid on canine and equine adipose-derived mesenchymal stem cells (ASCs) an in vitro research.
Nicpon J, et al.
Polish Journal of Veterinary Sciences, 18(1), 3-11 (2015)
Richard L Hornsby et al.
Scientific reports, 10(1), 9620-9620 (2020-06-17)
The causative agent of leptospirosis includes multiple serovars and species of pathogenic leptospires that are excreted via urine from reservoir hosts of infection. Primary isolation takes weeks to months, and is limited to semi-solid media at 28-30 °C. Here we present
Tobias M Lindig et al.
Histochemistry and cell biology, 131(1), 1-12 (2008-09-23)
We have compared the three-dimensional (3D) morphology of stubby and spiny neurons derived from the human small intestine. After immunohistochemical triple staining for leu-enkephalin (ENK), vasoactive intestinal peptide (VIP) and neurofilament (NF), neurons were selected and scanned based on their

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service