Skip to Content
Merck
All Photos(1)

Key Documents

MTOXCENULL

Sigma-Aldrich

CypExpress Control

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€263.00
10 G
€2,230.00

€263.00


Check Cart for Availability


Select a Size

Change View
1 G
€263.00
10 G
€2,230.00

About This Item

EC Number:
UNSPSC Code:
41106200
NACRES:
NA.77

€263.00


Check Cart for Availability

biological source

yeast

form

dry powder

shipped in

dry ice

storage temp.

−70°C

Biochem/physiol Actions

Cytochrome P450 is a heterogeneous family of isozymes whose primary function is to oxidize small molecules, both as a function of intermediary metabolism (e.g., fatty acids) and to detoxify exogenous compounds (drugs or toxins). Some isoforms have narrow substrate specificity, while others are promiscuous.

Features and Benefits

CypExpress Control is a permeabilized and stabilized dried yeast powder preparation using an empty vector and processed identically to CypExpress P450 products.

Physical form

Light yellow, very fine powder

Reconstitution

Per Protocol/Variable

Legal Information

6538 is a trademark of American Type Culture Collection
CypExpress is a trademark of Oxford Biomedical Research

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service