Skip to Content
Merck
All Photos(1)

Key Documents

MSST0058

Sigma-Aldrich

SILuLite IL8, Interleukin-8 human

recombinant, expressed in HEK 293 cells, MS Protein Standard

Synonym(s):

C-X-C motif chemokine 8, Chemokine (C-X-C motif), Emoctakin, Granulocyte chemotactic protein 1 (GCP-1), IL-8, Monocyte-derived neutrophil, Monocyte-derived neutrophil-activating peptide (MONAP), Neutrophil-activating protein 1 (NAP-1), Protein 3-10C, T-cell chemotactic factor, chemotactic factor (MDNCF), ligand 8

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€688.00

€688.00


Estimated to ship on07 May 2025


Request a Bulk Order

Select a Size

Change View
50 μG
€688.00

About This Item

UNSPSC Code:
23201100
NACRES:
NA.32

€688.00


Estimated to ship on07 May 2025


Request a Bulk Order

recombinant

expressed in HEK 293 cells

Quality Level

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

suitability

suitable for mass spectrometry (internal calibrator)

UniProt accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

human ... CXCL8(3576)

Related Categories

General description

SILuLite IL8 is a recombinant human protein expressed in human 293 cells. It is a mixture of the 3 main CXCL8 isoforms with the molecular weights, amino acid number and relative abundance as described in Table 1 of the product datasheet. SILuLite IL8 is an analytical standard designed to be used as starting material for preparation of calibrators and controls in LC-MS applications.

Biochem/physiol Actions

Interleukin-8 (IL-8) is a member of the CXC chemokine subfamily and is produced by blood cells and many types of tissues. The measurement of IL-8 in voided urinary samples may have utility for urine-based detection of bladder cancer. Urinary IL-8 was a strong biomarker of stress under intensive and prolonged demands, both acutely and over time. IL-8 and cathepsin B levels were significantly elevated in melanoma patients, and more importantly, the combination of IL-8 and cathepsin B were also studied as a prognosis marker for melanoma mortality.

Sequence

EGAVLPRSAKELRCQCIKTYSKPFHPKFIKELRVIESGPHCANTEIIVKLSDGRELCLDPKENWVQRVVEKFLKRAENS

Physical form

Supplied as a lyophilized powder containing phosphate buffered saline.

Legal Information

SILu is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service