Skip to Content
Merck
All Photos(9)

Key Documents

CS0006

Sigma-Aldrich

Soluble Collagen Quantification Assay Kit

Synonym(s):

Collagen Assay Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€605.00

€605.00


Check Cart for Availability


Select a Size

Change View
1 KIT
€605.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.84

€605.00


Check Cart for Availability

usage

sufficient for 200 fluorometric tests

shipped in

dry ice

storage temp.

−20°C

General description

Collagen is one of the most abundant proteins in connective tissues and internal organs of mammals. Collagen, a structural component, provides the tensile strength of the extracellular matrix (ECM) and is classified into several structurally and genetically distinct types. Although different types of collagen exist, they are all composed of molecules with three polypeptide chains that are arranged in a triple helical conformation. Slight differences in the primary structure (amino acid sequence) establish differences between the types.
The Soluble Collagen Quantification Assay Kit provides a simple and sensitive procedure for measuring soluble collagen in various sample types.
One of the advantages using our soluble collagen assay kit is that it does not require the use of the hazardous and corrosive perchloric acid as oppose to other kits in the market. Thus, it does not require a cumbersome and time-consuming procedure. You will get the result within 60 minutes: The amount of soluble collagen is determined based on an enzymatic reaction, where collagen is specifically digested into peptides. Subsequently, the collagen peptides are labeled with a fluorescent probe. The fluorescence intensity, measured at λ ex = 375 nm/ λ em = 465 nm, is proportional to the amount of soluble collagen in the sample.
This kit can be used to quantify soluble collagen extracted from soft tissues (e.g., muscle, heart, etc.), tissue culture cell lysate and medium, serum samples, collagen in food, and purified collagens of various sources. This kit can detect purified collagen types I, II, III, IV and V.

The detection range of the kit is 0.2-2 μg of soluble collagen.

Application

Measuring soluble collagen in various sample types

Features and Benefits

Our economical Soluble Collagen Quantification Assay Kit also saves your time and efforts:

  • Highly sensitive, simple and high-throughput assay for measuring soluble collagen in various samples
  • User friendly: No need to weigh or calculate needed amounts of reagents on your own
  • Customer safety: No need to use hazardous materials like strong acids, unlike other kits on the market which are based on hydroxyproline analysis.
  • Supportive calculator (Click here to download a calculator excel file):Will calculate the reagents needed according to your experimental needs, and analyses your results based on your experimental data.
  • Quick instruction bench card - to assure your experimental success
  • More experiments in one kit - contains enough reagents for 200 tests.
  • Detection time: only 60 minutes

Suitability

Suitable for the detection of Soluble Collagen various biological samples and cultured cell medium.

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dongxu Feng et al.
The Journal of biological chemistry, 295(39), 13640-13650 (2020-08-05)
Progranulin (PGRN) is an autocrine growth factor that exerts crucial roles within cartilage tissue; however, the molecular mechanisms underlying PGRN-mediated cartilage homeostasis remain elusive. In the present study, we investigated the role of PGRN in regulating chondrocyte homeostasis and its

Questions

1–2 of 2 Questions  
  1. Whether soluble collagen quantification assay KIT (CS0006) can test the collagen expressed from mouse L929 cells?

    1 answer
    1. We have not tested collagen expressed in L929 cells, but we expect it to work.

      Helpful?

  2. Can this kit quantify soluble collagen peptides that don't possess hydroxyprolines?

    1 answer
    1. Yes, this product will be suitable. The mechanism is not dependent on the presence of hydroxyproline.

      Please see the link below to review the product datasheet:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/239/884/cs0006pis-ms.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service