Skip to Content
Merck
All Photos(1)

Key Documents

MSST0030

Sigma-Aldrich

SILuLite APOA2 Apolipoprotein A-II human

recombinant, expressed in HEK 293 cells, MS Protein Standard

Synonym(s):

Apo-AII human, ApoA-II, ApolipoproteinA2

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€687.00

€687.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 μG
€687.00

About This Item

UNSPSC Code:
23201100
NACRES:
NA.12

€687.00


Check Cart for Availability

Request a Bulk Order

biological source

human

Quality Level

recombinant

expressed in HEK 293 cells

Assay

≥98% (SDS-PAGE)

form

lyophilized powder

potency

≥98 Heavy amino acids incorporation efficiency by MS

technique(s)

mass spectrometry (MS): suitable

suitability

suitable for mass spectrometry (internal calibrator)

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... APOA2(336)

Related Categories

General description

Apolipoprotein A-II (ApoA-II) occurs in plasma as a dimer of two 77-amino acid chains linked by a disulfide bridge. After apoA-I, it is the second major protein component of HDL, accounting for approximately 20% of HDL total protein. ApoA-II is thought to play an important role in triglyceride metabolism both from animal and human studies. 3 Recent findings attribute apoA-II to inhibitory effects on lipoprotein lipase-mediated hydrolysis of triglyceride-rich particles. Additional associations of apoA-II have been reported for a variety of protein factors including hepatic lipase (HL), lipoprotein lipase (LPL), endothelial lipase, CETP, PLTP, and LCAT.

Immunogen

MAEPRQEFEVMEDHAGTYGLGDRKDQGGYTMHQDQEGDTDAGLKESPLQTPTEDGSEEPGSETSDAKSTPTAEDVTAPLVDEGAPGKQAAAQPHTEIPEGTTAEEAGIGDTPSLEDEAAGHVTQARMVSKSKDGTGSDDKKAKGADGKTKIATPRGAAPPGQKGQANATRIPAKTPPAPKTPPSSGEPPKSGDRSGYSSPGSPGTPGSRSRTPSLPTPPTREPKKVAVVRTPPKSPSSAKSRLQTAPVPMPDLKNVKSKIGSTENLKHQPGGGKVQIINKKLDLSNVQSKCGSKDNIKHVPGGGSVQIVYKPVDLSKVTSKCGSLGNIHHKPGGGQVEVKSEKLDFKDRVQSKIGSLDNITHVPGGGNKKIETHKLTFRENAKAKTDHGAEIVYKSPVVSGDTSPRHLSNVSSTGSIDMVDSPQLATLADEVSASLAKQGLDRIRGRKLGPFEGKPIPNPLLGLDSTRTGHHHHHHHHGGQ

Biochem/physiol Actions

SILuLite APOA2 is a recombinant human protein expressed in human 293 cells. It is a homodimer consisting of 97 amino acids (including C-terminal polyhistidine and flag tags), with a calculated molecular mass of 11 kDa. SILu Lite APOA2 is an analytical standard designed to be used as starting material for preparation of calibrators and controls in LC-MS applications.

Physical form

Supplied as a lyophilized powder containing phosphate buffered saline.

Legal Information

SILu is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service