Skip to Content
Merck
All Photos(1)

Documents

67649

Supelco

Methylmalonic acid

analytical standard

Synonym(s):

2-Methylpropanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(CO2H)2
CAS Number:
Molecular Weight:
118.09
Beilstein:
1756084
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Assay

≥98.5% (GC)

form

solid

shelf life

limited shelf life, expiry date on the label

mp

128-130 °C (lit.)
131-134 °C

SMILES string

CC(C(O)=O)C(O)=O

InChI

1S/C4H6O4/c1-2(3(5)6)4(7)8/h2H,1H3,(H,5,6)(H,7,8)

InChI key

ZIYVHBGGAOATLY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service