Skip to Content
Merck
All Photos(1)

Key Documents

MSST0014

Sigma-Aldrich

SILuLite AMBP Alpha-1 Microglycoprotein human

recombinant, expressed in HEK 293 cells, MS protein standard

Synonym(s):

Alpha-1-microglobulin, Complex-forming glycoprotein heterogeneous in charge

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€708.00

€708.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
50 μG
€708.00

About This Item

UNSPSC Code:
23201100
NACRES:
NA.12

€708.00


Estimated to ship on25 May 2025


Request a Bulk Order

biological source

human

Quality Level

recombinant

expressed in HEK 293 cells

tag

His tagged

Assay

≥98% (SDS-PAGE)

form

lyophilized powder

technique(s)

mass spectrometry (MS): suitable

suitability

suitable for mass spectrometry (internal calibrator)

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... AMBP(259)

Related Categories

General description

SILu Lite AMBP is a recombinant human protein expressed in human 293 cells. It is a monomer of 204 amino acids (including C-terminal polyhistidine and flag tags), with a molecular weight of ~23 kDa. SILu Lite AMBP is an analytical standard designed to be used as starting material for preparation of calibrators and controls in LC-MS applications.

Biochem/physiol Actions

AMBP is synthesized by the liver. Approximately half of the circulating protein is complexed to IgA. The free form of AMBP is filtered by the glomerulus and reabsorbed by proximal tubule cells.[1] AMBP has been found to be a sensitive biomarker for proximal tubular dysfunction even in the early phase of injury when no histologic damage is observable. In addition, urinary AMBP has been proposed to be a useful marker of tubular dysfunction even in low-gestational-age preterm infants, a population at high risk for AKI (Acute Kidney Injury). Urinary AMBP is also a marker of kidney damage in type 2 diabetes.[2]

Sequence

GPVPTPPDNIQVQENFNISRIYGKWYNLAIGSTCPWLKKIMDRMTVSTLVLGEGATEAEISMTSTRWRKGVCEETSGAYEKTDTDGKFLYHKSKWNITMESYVVHTNYDEYAIFLTKKFSRHHGPTITAKLYGRAPQLRETLLQDFRVVAQGVGIPEDSIFTMADRGECVPGEQEPEPILIPRVDYKDDDDKGHHHHHHHHGGQ

Physical form

Supplied as a lyophilized powder containing phosphate buffered saline.

Legal Information

This product is licensed under U.S. Patent No. 7,396,688 and foreign counterparts from E. I. du Pont de Nemours and Company. The purchase of this product conveys to the buyer the nontransferable right to use the purchased amount of the product for research and development only, including services for a third party for consideration. The buyer cannot sell or otherwise transfer this product, its components or materials made using this product or its components to a third party. Information about licenses for excluded uses is available from: E. I. du Pont de Nemours and Company; Attn: Associate Director, Commercial Development; DuPont Experimental Station E268; 200 Powdermill Rd.; Wilmington, DE 19803; 1-877-881-9787 (voice), 1-302-695-1437 (fax), [email protected].
SILu is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elisa Bellei et al.
The journal of headache and pain, 16, 559-559 (2015-08-15)
Medication-overuse headache (MOH) is a chronic disorder that results from the overuse of analgesics drugs, triptans or other acute headache compounds. Although the exact mechanisms underlying MOH remain still unknown, several studies suggest that it may be associated with development
Vishal S Vaidya et al.
Annual review of pharmacology and toxicology, 48, 463-493 (2007-10-17)
Acute kidney injury (AKI) is a common condition with a high risk of death. The standard metrics used to define and monitor the progression of AKI, such as serum creatinine and blood urea nitrogen levels, are insensitive, nonspecific, and change

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service