Skip to Content
Merck
All Photos(2)

Key Documents

MSST0012

Sigma-Aldrich

SILuLite ALB Albumin human

recombinant, expressed in HEK 293 cells, MS Protein Standard

Synonym(s):

Albumin, Mass spectrometry standard, Human Albumin

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€708.00

€708.00


Check Cart for Availability


Select a Size

Change View
50 μG
€708.00

About This Item

UNSPSC Code:
23201100
NACRES:
NA.12

€708.00


Check Cart for Availability

biological source

human

Quality Level

recombinant

expressed in HEK 293 cells

Assay

≥98% (SDS-PAGE)

form

lyophilized powder

technique(s)

mass spectrometry (MS): suitable

suitability

suitable for mass spectrometry (internal calibrator)

UniProt accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

human ... ALB(213)

General description

SILuLite ALB is a recombinant human protein expressed in human 293 cells. It is a monomer of 607 amino acids (including C-terminal polyhistidine and flag tags), with a molecular weight of ~69 kDa. SILuLite ALB is an analytical standard designed to be used as starting material for preparation of calibrators and controls in LC-MS applications.

Biochem/physiol Actions

Albumin is synthesized exclusively by the liver parenchymal cells at a rate of 14 g/day. While it is the most abundant plasma protein (>50%), increased urine albumin concentrations indicate bleedings in the lower urinary tract (ureter, bladder) or infections of the renal pelvis. A small abnormal albumin excretion is known as Microalbuminuria and serves as an indicator for temporary overload of the glomerular filtration (fever, excessive sports) or chronic injury of the glomeruli (diabetes).[1][2]

Sequence

RRDAHKSEVAHRFKDLGEENFKALVLIAFAQYLQQCPFEDHVKLVNEVTEFAKTCVADESAENCDKSLHTLFGDKLCTVATLRETYGEMADCCAKQEPERNECFLQHKDDNPNLPRLVRPEVDVMCTAFHDNEETFLKKYLYEIARRHPYFYAPELLFFAKRYKAAFTECCQAADKAACLLPKLDELRDEGKASSAKQRLKCASLQKFGERAFKAWAVARLSQRFPKAEFAEVSKLVTDLTKVHTECCHGDLLECADDRADLAKYICENQDSISSKLKECCEKPLLEKSHCIAEVENDEMPADLPSLAADFVESKDVCKNYAEAKDVFLGMFLYEYARRHPDYSVVLLLRLAKTYETTLEKCCAAADPHECYAKVFDEFKPLVEEPQNLIKQNCELFEQLGEYKFQNALLVRYTKKVPQVSTPTLVEVSRNLGKVGSKCCKHPEAKRMPCAEDYLSVVLNQLCVLHEKTPVSDRVTKCCTESLVNRRPCFSALEVDETYVPKEFNAETFTFHADICTLSEKERQIKKQTALVELVKHKPKATKEQLKAVMDDFAAFVEKCCKADDKETCFAEEGKKLVAASQAALGLDYKDDDDKGHHHHHHHHGGQ

Physical form

Supplied as a lyophilized powder containing phosphate buffered saline.

Legal Information

This product is licensed under U.S. Patent No. 7,396,688 and foreign counterparts from E. I. du Pont de Nemours and Company. The purchase of this product conveys to the buyer the nontransferable right to use the purchased amount of the product for research and development only, including services for a third party for consideration. The buyer cannot sell or otherwise transfer this product, its components or materials made using this product or its components to a third party. Information about licenses for excluded uses is available from: E. I. du Pont de Nemours and Company; Attn: Associate Director, Commercial Development; DuPont Experimental Station E268; 200 Powdermill Rd.; Wilmington, DE 19803; 1-877-881-9787 (voice), 1-302-695-1437 (fax), [email protected].
SILu is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lorraine B Ware et al.
American journal of physiology. Renal physiology, 300(3), F628-F638 (2010-12-15)
This study evaluated the potential utility of albuminuria as a "biomarker" of acute kidney injury (AKI) and tested whether AKI induces renal expression of the normally silent albumin gene. Urine albumin concentrations were measured in mice with five different AKI
Salvador Lopez-Giacoman et al.
World journal of nephrology, 4(1), 57-73 (2015-02-11)
Chronic kidney disease (CKD) typically evolves over many years, with a long latent period when the disease is clinically silent and therefore diagnosis, evaluation and treatment is based mainly on biomarkers that assess kidney function. Glomerular filtration rate (GFR) remains

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service